Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu479235_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-30e-5p
          Stem-loop Accession Number: MI0000749
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 40754355 - 40754446 [+] on Build GRCh38
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Species: Human, Mouse, Rat, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu479235_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          UGUAAACAUCCUUGACUGGAAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30e-5p MIMAT0000692
          Mouse mmu-miR-30e-5p MIMAT0000248
          Rat rno-miR-30e-5p MIMAT0000805
          Horse eca-miR-30e MIMAT0012916
          Pan troglodytes ptr-miR-30e MIMAT0008092
          Pongo pygmaeus ppy-miR-30e MIMAT0015739
          View More Rows
          Rhesus monkey mml-miR-30e-5p MIMAT0006172
          Xenopus tropicalis xtr-miR-30e MIMAT0003678
          Zebrafish dre-miR-30e-5p MIMAT0001807

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30e
          Stem-loop Accession # MI0000749
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 1 - 40754355 - 40754446 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000692
          miRBase ID: hsa-miR-30e-5p
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: Chr. 1 - 40754355 - 40754446 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30e
          Stem-loop Accession # MI0000259
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUGAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 4 - 120772606 - 120772697 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000248
          miRBase ID: mmu-miR-30e-5p
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: Chr. 4 - 120772606 - 120772697 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30e
          Stem-loop Accession # MI0000867
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUGAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 5 - 139702872 - 139702963 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000805
          miRBase ID: rno-miR-30e-5p
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: Chr. 5 - 139702872 - 139702963 [-] on Build Rnor_6.0

          Horse

          Stem-loop ID eca-mir-30e
          Stem-loop Accession # MI0012672
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012916
          miRBase ID: eca-miR-30e
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-30e
          Stem-loop Accession # MI0008606
          Stem-loop Sequence
          GGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008092
          miRBase ID: ptr-miR-30e
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-30e
          Stem-loop Accession # MI0014805
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015739
          miRBase ID: ppy-miR-30e
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-30e
          Stem-loop Accession # MI0007602
          Stem-loop Sequence
          GGGCAGUCUUCGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006172
          miRBase ID: mml-miR-30e-5p
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-30e
          Stem-loop Accession # MI0004920
          Stem-loop Sequence
          GGGCAGUUACAGCCUCUGUAAACAUCCUUGACUGGAAGCUGUGAAAUAAUGAUAGUAGCUUUCAGUCGGAUGUUUACAGCUGCUAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003678
          miRBase ID: xtr-miR-30e
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-30e-2
          Stem-loop Accession # MI0001950
          Stem-loop Sequence
          UACGGGCUACUGUAAACAUCCUUGACUGGAAGCUGGUGCACAUGAUGGAGCUUUCAGUCGGAUGUUUGCAGCAGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001807
          miRBase ID: dre-miR-30e-5p
          Mature miRNA Sequence: UGUAAACAUCCUUGACUGGAAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303424_pri
          TaqMan™ Pri-miRNA Assay : Rn03464782_pri
          TaqMan™ Pri-miRNA Assay : Mm03306569_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10037
          Ambion® Pre-miR™ miRNA Precursor : PM10037
          TaqMan™ MicroRNA Assay : 002223
          mirVana® miRNA inhibitor : MH10037
          mirVana® miRNA mimic : MC10037
          TaqMan™ Advanced miRNA Assay : rno479235_mir
          TaqMan™ Advanced miRNA Assay : 479235_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline