Search
Search

| Assay Name: | hsa-miR-30e-5p |
|---|---|
| miRBase Accession Number: | MI0000749 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUAAACAUCCUUGACUGGAAG |
| Species: | Human, Mouse, Rat, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis, Zebrafish |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUUGACUGGAAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30e-5p | MIMAT0000692 |
| Mouse | mmu-miR-30e-5p | MIMAT0000248 |
| Rat | rno-miR-30e-5p | MIMAT0000805 |
| Horse | eca-miR-30e | MIMAT0012916 |
| Pan troglodytes | ptr-miR-30e | MIMAT0008092 |
| Pongo pygmaeus | ppy-miR-30e | MIMAT0015739 |
| View More Rows | ||
| Rhesus monkey | mml-miR-30e-5p | MIMAT0006172 |
| Xenopus tropicalis | xtr-miR-30e | MIMAT0003678 |
| Zebrafish | dre-miR-30e-5p | MIMAT0001807 |