Search
Search

| Assay Name: | mmu-miR-181d-5p |
|---|---|
| Stem-loop Accession Number: | MI0003139 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACAUUCAUUGUUGUCGGUGGGU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Gorilla gorilla, Otolemur garnettii, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAUUGUUGUCGGUGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-181d-5p | MIMAT0002821 |
| Mouse | mmu-miR-181d-5p | MIMAT0004324 |
| Rat | rno-miR-181d-5p | MIMAT0005299 |
| Bovine | bta-miR-181d | MIMAT0009243 |
| Callithrix jacchus | cja-miR-181d | MIMAT0039347 |
| Cricetulus griseus | cgr-miR-181d-5p | MIMAT0023803 |
| View More Rows | ||
| Dog | cfa-miR-181d | MIMAT0006636 |
| Goat | chi-miR-181d | MIMAT0036005 |
| Gorilla gorilla | ggo-miR-181d | MIMAT0024100 |
| Otolemur garnettii | oga-miR-181d | MIMAT0049710 |
| Pan troglodytes | ptr-miR-181d | MIMAT0008049 |
| Pongo pygmaeus | ppy-miR-181d | MIMAT0015779 |
| Rhesus monkey | mml-miR-181d | MIMAT0006217 |
| Tupaia chinensis | tch-miR-181d | MIMAT0036528 |