Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno479517_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-181d-5p
          Stem-loop Accession Number: MI0003139
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 13874875 - 13875011 [+] on Build GRCh38
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Gorilla gorilla, Otolemur garnettii, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno479517_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAUUGUUGUCGGUGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181d-5p MIMAT0002821
          Mouse mmu-miR-181d-5p MIMAT0004324
          Rat rno-miR-181d-5p MIMAT0005299
          Bovine bta-miR-181d MIMAT0009243
          Callithrix jacchus cja-miR-181d MIMAT0039347
          Cricetulus griseus cgr-miR-181d-5p MIMAT0023803
          View More Rows
          Dog cfa-miR-181d MIMAT0006636
          Goat chi-miR-181d MIMAT0036005
          Gorilla gorilla ggo-miR-181d MIMAT0024100
          Otolemur garnettii oga-miR-181d MIMAT0049710
          Pan troglodytes ptr-miR-181d MIMAT0008049
          Pongo pygmaeus ppy-miR-181d MIMAT0015779
          Rhesus monkey mml-miR-181d MIMAT0006217
          Tupaia chinensis tch-miR-181d MIMAT0036528

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181d
          Stem-loop Accession # MI0003139
          Stem-loop Sequence
          GUCCCCUCCCCUAGGCCACAGCCGAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACUGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACACGGCUUAAGGGGAAUGGGGAC
          Chromosome Location Chr. 19 - 13874875 - 13875011 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002821
          miRBase ID: hsa-miR-181d-5p
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: Chr. 19 - 13874875 - 13875011 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-181d
          Stem-loop Accession # MI0005450
          Stem-loop Sequence
          ACAAUUAACAUUCAUUGUUGUCGGUGGGUUGUGAGGAGGCAGCCAGACCCACCGGGGGAUGAAUGUCACUGU
          Chromosome Location Chr. 8 - 84178716 - 84178787 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004324
          miRBase ID: mmu-miR-181d-5p
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: Chr. 8 - 84178716 - 84178787 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-181d
          Stem-loop Accession # MI0006132
          Stem-loop Sequence
          GGUCACAAUUAACAUUCAUUGUUGUCGGUGGGUUGUGAGGAGAAGACCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGG
          Chromosome Location Chr. 19 - 25290051 - 25290133 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005299
          miRBase ID: rno-miR-181d-5p
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: Chr. 19 - 25290051 - 25290133 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-181d
          Stem-loop Accession # MI0009754
          Stem-loop Sequence
          UCCCCUCCCUUAGGCCACAGCGAAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGACAGAGACCAGACCCACCGAGGGAUGAAUGUCACUGUGGCUGGGCCAGACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009243
          miRBase ID: bta-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-181d
          Stem-loop Accession # MI0031862
          Stem-loop Sequence
          CCCCCUCCCCUAAGCCACAGCCAAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACAUGGCUUAAGGGGAAUAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039347
          miRBase ID: cja-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-181d
          Stem-loop Accession # MI0020423
          Stem-loop Sequence
          GUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAUGAGGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023803
          miRBase ID: cgr-miR-181d-5p
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-181d
          Stem-loop Accession # MI0008035
          Stem-loop Sequence
          AACAUUCAUUGUUGUCGGUGGGUUGUGAGGACGGAGGCCAGACCCACCGAGGGAUGAAUGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006636
          miRBase ID: cfa-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-181d
          Stem-loop Accession # MI0030652
          Stem-loop Sequence
          UCCCCUCCCUUAGGCCACAGCAAAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGACGGAGACCAGACCCACCGAGGGAUGAAUGUCACUGUGGCUGGGCCAGACACAGCCUCGGGGGAAUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036005
          miRBase ID: chi-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-181d
          Stem-loop Accession # MI0020647
          Stem-loop Sequence
          CCACAGCCGAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACACGGCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024100
          miRBase ID: ggo-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-181d
          Stem-loop Accession # MI0040496
          Stem-loop Sequence
          AACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCAAGGCCAGACCCACCGGGGGAUGAAUGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049710
          miRBase ID: oga-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-181d
          Stem-loop Accession # MI0008557
          Stem-loop Sequence
          UCCCCUCCCCUAGGCCACAGCCGAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACACGGCUUAAGGGGAAUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008049
          miRBase ID: ptr-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-181d
          Stem-loop Accession # MI0014847
          Stem-loop Sequence
          UCCCCUCCCCUAGGCCACAGCCAAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACACGGCUUAAGGGGAAUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015779
          miRBase ID: ppy-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-181d
          Stem-loop Accession # MI0007647
          Stem-loop Sequence
          GUCCCCUCCCCUAGGCCACAGCCAAGGUCACAAUCAACAUUCAUUGUUGUCGGUGGGUUGUGAGGACCGAGGCCAGACCCACCGGGGGAUGAAUGUCACUGUGGCUGGGCCAGACACGGCUUAAGGGGAAUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006217
          miRBase ID: mml-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-181d
          Stem-loop Accession # MI0031187
          Stem-loop Sequence
          AACAUUCAUUGUUGUCGGUGGGUUGUGAGGACGGAGCCCAGACCCACCGGGGGAUGAAUGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036528
          miRBase ID: tch-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGUUGUCGGUGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466046_pri
          TaqMan™ Pri-miRNA Assay : Hs03303910_pri
          TaqMan™ Pri-miRNA Assay : Mm03307266_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12522
          Ambion® Pre-miR™ miRNA Precursor : PM12522
          mirVana® miRNA inhibitor : MH12522
          mirVana® miRNA mimic : MC12522
          TaqMan™ Advanced miRNA Assay : mmu479517_mir
          TaqMan™ Advanced miRNA Assay : 479517_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline