Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480905_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-1249-3p
          Stem-loop Accession Number: MI0006384
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 45200954 - 45201019 [-] on Build GRCh38
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Dog, Eptesicus fuscus, Goat, Gorilla gorilla, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480905_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000667, mir-1249

          Mature miRNA Sequence:

          ACGCCCUUCCCCCCCUUCUUCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1249-3p MIMAT0005901
          Mouse mmu-miR-1249-3p MIMAT0010560
          Rat rno-miR-1249 MIMAT0017892
          Bovine bta-miR-1249 MIMAT0009976
          Cricetulus griseus cgr-miR-1249 MIMAT0041147
          Dasypus novemcinctus dno-miR-1249-3p MIMAT0048032
          View More Rows
          Dog cfa-miR-1249 MIMAT0034321
          Eptesicus fuscus efu-miR-1249 MIMAT0034894
          Goat chi-miR-1249 MIMAT0035921
          Gorilla gorilla ggo-miR-1249 MIMAT0024247
          Pan troglodytes ptr-miR-1249 MIMAT0007975
          Pig ssc-miR-1249 MIMAT0025385
          Pongo pygmaeus ppy-miR-1249 MIMAT0016174
          Rabbit ocu-miR-1249-3p MIMAT0048586
          Rhesus monkey mml-miR-1249 MIMAT0024324

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1249
          Stem-loop Accession # MI0006384
          Stem-loop Sequence
          GGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGGCUGGAACGCCCUUCCCCCCCUUCUUCACCUG
          Chromosome Location Chr. 22 - 45200954 - 45201019 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005901
          miRBase ID: hsa-miR-1249-3p
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: Chr. 22 - 45200954 - 45201019 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-1249
          Stem-loop Accession # MI0004132
          Stem-loop Sequence
          CGAUUCUUGUUUGCGUGGGGAGGAGGGAGGGGAUGGGCCAAGUUCCCUAUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCG
          Chromosome Location Chr. 15 - 84951526 - 84951623 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0010560
          miRBase ID: mmu-miR-1249-3p
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: Chr. 15 - 84951526 - 84951623 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-1249
          Stem-loop Accession # MI0015470
          Stem-loop Sequence
          UCUUGCUGCUCCGAUUCUUGUUUGCAUGGGGAGGAGGGAGGGGAUGGGCCAAGUUCCCUAUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCGUCCUUCAAGAC
          Chromosome Location Chr. 7 - 125861192 - 125861311 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017892
          miRBase ID: rno-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: Chr. 7 - 125861192 - 125861311 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-1249
          Stem-loop Accession # MI0010481
          Stem-loop Sequence
          GGGAGGAGGGAGGAGAUGGGCCACGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009976
          miRBase ID: bta-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-1249
          Stem-loop Accession # MI0033076
          Stem-loop Sequence
          CGAUUCUUGUUUGCAUGGGGAGGAGGGAGGGGAUGGGCCAAGUUCCCUGUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041147
          miRBase ID: cgr-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-1249
          Stem-loop Accession # MI0039183
          Stem-loop Sequence
          AGGAGGGAGGAGAGGGGCCAAGUUCCCGCUGCCUGGAACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048032
          miRBase ID: dno-miR-1249-3p
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-1249
          Stem-loop Accession # MI0027912
          Stem-loop Sequence
          UCCCAGCUCGCCUUGUCCUCAGAUUCUUGUUUGCAUGGGGAGGAGGGAGGAGAGGGGCCAAGUUCCCUCCGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCGUCCUUCAAGACCCAGCUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0034321
          miRBase ID: cfa-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-1249
          Stem-loop Accession # MI0028578
          Stem-loop Sequence
          UUUGCAUGGGGAGGAGGGAGGAGAGGGGCCACGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0034894
          miRBase ID: efu-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-1249
          Stem-loop Accession # MI0030603
          Stem-loop Sequence
          UCCUCAGAUUCUUGUUUGCAUGGGGAGGAGGGAGGAGAUGGGCCACGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUAGCGAACUCCUACUCGUCCUUCAAGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035921
          miRBase ID: chi-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-1249
          Stem-loop Accession # MI0020794
          Stem-loop Sequence
          CAUCCUGCUCCUCAGAUUAUUGUUGGCAUGGGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGGCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024247
          miRBase ID: ggo-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-1249
          Stem-loop Accession # MI0008455
          Stem-loop Sequence
          GGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGGCUGGAACGCCCUUCCCCCCCUUCUUCACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007975
          miRBase ID: ptr-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-1249-1
          Stem-loop Accession # MI0022158
          Stem-loop Sequence
          UGUUUGCAUGGGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUAGCGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025385
          miRBase ID: ssc-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Stem-loop ID ssc-mir-1249-2
          Stem-loop Accession # MI0022159
          Stem-loop Sequence
          UGUUUGCAUGGGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCACCUAGCGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025385
          miRBase ID: ssc-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-1249
          Stem-loop Accession # MI0015207
          Stem-loop Sequence
          GGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGGCUGGAACGCCCUUCCCCCCCUUCUUCACCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016174
          miRBase ID: ppy-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-1249
          Stem-loop Accession # MI0039475
          Stem-loop Sequence
          AGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGCCUGGAACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048586
          miRBase ID: ocu-miR-1249-3p
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1249
          Stem-loop Accession # MI0020872
          Stem-loop Sequence
          CAUCCUGCUCCUCAGAUUCUUGUUGGCAUGGGGAGGAGGGAGGAGAUGGGCCAAGUUCCCUCUGGCUGGAACGCCCUUCCCCCCCUUCUUCACCUGGCGAACUCCUACUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024324
          miRBase ID: mml-miR-1249
          Mature miRNA Sequence: ACGCCCUUCCCCCCCUUCUUCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305741_pri
          TaqMan™ Pri-miRNA Assay : Rn04229838_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13203
          Ambion® Pre-miR™ miRNA Precursor : PM13203
          TaqMan™ MicroRNA Assay : 002868
          mirVana® miRNA inhibitor : MH13203
          mirVana® miRNA mimic : MC13203
          TaqMan™ Advanced miRNA Assay : rno480905_mir
          TaqMan™ Advanced miRNA Assay : 478654_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline