Search
Search

| Assay Name: | hsa-miR-1249-3p |
|---|---|
| miRBase Accession Number: | MI0006384 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACGCCCUUCCCCCCCUUCUUCA |
| Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Dog, Eptesicus fuscus, Goat, Gorilla gorilla, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000667, mir-1249 |
Mature miRNA Sequence: | ACGCCCUUCCCCCCCUUCUUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1249-3p | MIMAT0005901 |
| Mouse | mmu-miR-1249-3p | MIMAT0010560 |
| Rat | rno-miR-1249 | MIMAT0017892 |
| Bovine | bta-miR-1249 | MIMAT0009976 |
| Cricetulus griseus | cgr-miR-1249 | MIMAT0041147 |
| Dasypus novemcinctus | dno-miR-1249-3p | MIMAT0048032 |
| View More Rows | ||
| Dog | cfa-miR-1249 | MIMAT0034321 |
| Eptesicus fuscus | efu-miR-1249 | MIMAT0034894 |
| Goat | chi-miR-1249 | MIMAT0035921 |
| Gorilla gorilla | ggo-miR-1249 | MIMAT0024247 |
| Pan troglodytes | ptr-miR-1249 | MIMAT0007975 |
| Pig | ssc-miR-1249 | MIMAT0025385 |
| Pongo pygmaeus | ppy-miR-1249 | MIMAT0016174 |
| Rabbit | ocu-miR-1249-3p | MIMAT0048586 |
| Rhesus monkey | mml-miR-1249 | MIMAT0024324 |