Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480934_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-142a-3p
          Stem-loop Accession Number: MI0000458
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 58331232 - 58331318 [-] on Build GRCh38
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Artibeus jamaicensis, Eptesicus fuscus, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480934_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000084, mir-142

          Mature miRNA Sequence:

          UGUAGUGUUUCCUACUUUAUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-142-3p MIMAT0000434
          Mouse mmu-miR-142a-3p MIMAT0000155
          Rat rno-miR-142-3p MIMAT0000848
          Alligator mississippiensis ami-miR-142-3p MIMAT0038163
          Artibeus jamaicensis aja-miR-142 MIMAT0031136
          Eptesicus fuscus efu-miR-142 MIMAT0035200
          View More Rows
          Horse eca-miR-142-3p MIMAT0013023
          Monodelphis domestica mdo-miR-142 MIMAT0004112
          Ophiophagus hannah oha-miR-142-3p MIMAT0036722
          Pan troglodytes ptr-miR-142 MIMAT0008037
          Pongo pygmaeus ppy-miR-142-3p MIMAT0015766
          Rhesus monkey mml-miR-142-3p MIMAT0006200
          Xenopus laevis xla-miR-142-3p MIMAT0046455
          Xenopus tropicalis xtr-miR-142-3p MIMAT0003603
          Zebrafish dre-miR-142a-3p MIMAT0003160

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-142
          Stem-loop Accession # MI0000458
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000434
          miRBase ID: hsa-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-142a
          Stem-loop Accession # MI0000167
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000155
          miRBase ID: mmu-miR-142a-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-142
          Stem-loop Accession # MI0000915
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000848
          miRBase ID: rno-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-142-1
          Stem-loop Accession # MI0029746
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAAACAGCACUGCAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038163
          miRBase ID: ami-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Stem-loop ID ami-mir-142-2
          Stem-loop Accession # MI0029747
          Stem-loop Sequence
          CGGUGACAGUCUCCCCCCCCCAUAAAGUAGCGAGCACGACUCCGCCCCCGCGCCCGUGUAGUGUUUCCUACUUUAUGGAGGGGGUGGCUGGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038163
          miRBase ID: ami-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Artibeus jamaicensis

          Stem-loop ID aja-mir-142
          Stem-loop Accession # MI0024358
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031136
          miRBase ID: aja-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-142
          Stem-loop Accession # MI0028917
          Stem-loop Sequence
          UGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035200
          miRBase ID: efu-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-142
          Stem-loop Accession # MI0012771
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013023
          miRBase ID: eca-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-142
          Stem-loop Accession # MI0005301
          Stem-loop Sequence
          GCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCAAUGUAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004112
          miRBase ID: mdo-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-142
          Stem-loop Accession # MI0031355
          Stem-loop Sequence
          UAUGGACAGUGGACUCAUCCAUAAAGUAGAAAGCACUACUAAACGCUUGUGUCCAAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036722
          miRBase ID: oha-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-142
          Stem-loop Accession # MI0008543
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008037
          miRBase ID: ptr-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-142
          Stem-loop Accession # MI0014833
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015766
          miRBase ID: ppy-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-142
          Stem-loop Accession # MI0007631
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006200
          miRBase ID: mml-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-142-2
          Stem-loop Accession # MI0038253
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAGACAGCACUGAACGCUGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046455
          miRBase ID: xla-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-142-1
          Stem-loop Accession # MI0004842
          Stem-loop Sequence
          CAGUGCAGCCACCCAUAAAGUAGAAAGCACUACUAGACAGGACUGAACGCUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003603
          miRBase ID: xtr-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Stem-loop ID xtr-mir-142-2
          Stem-loop Accession # MI0004843
          Stem-loop Sequence
          CAGUGCAGCCACCCAUAAAGUAGAAAGCACUACUAGACAGGACUGAACGCUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003603
          miRBase ID: xtr-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-142a
          Stem-loop Accession # MI0002005
          Stem-loop Sequence
          CGUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACCCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003160
          miRBase ID: dre-miR-142a-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303162_pri
          TaqMan™ Pri-miRNA Assay : Rn03465064_pri
          TaqMan™ Pri-miRNA Assay : Mm03306331_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10398
          Ambion® Pre-miR™ miRNA Precursor : PM10398
          TaqMan™ MicroRNA Assay : 000464
          mirVana® miRNA inhibitor : MH10398
          mirVana® miRNA mimic : MC10398
          TaqMan™ Advanced miRNA Assay : 477910_mir
          TaqMan™ Advanced miRNA Assay : rno480934_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline