Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477910_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-142-3p
          Stem-loop Accession Number: MI0000458
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 58331232 - 58331318 [-] on Build GRCh38
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Artibeus jamaicensis, Eptesicus fuscus, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477910_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000084, mir-142

          Mature miRNA Sequence:

          UGUAGUGUUUCCUACUUUAUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-142-3p MIMAT0000434
          Mouse mmu-miR-142a-3p MIMAT0000155
          Rat rno-miR-142-3p MIMAT0000848
          Alligator mississippiensis ami-miR-142-3p MIMAT0038163
          Artibeus jamaicensis aja-miR-142 MIMAT0031136
          Eptesicus fuscus efu-miR-142 MIMAT0035200
          View More Rows
          Horse eca-miR-142-3p MIMAT0013023
          Monodelphis domestica mdo-miR-142 MIMAT0004112
          Ophiophagus hannah oha-miR-142-3p MIMAT0036722
          Pan troglodytes ptr-miR-142 MIMAT0008037
          Pongo pygmaeus ppy-miR-142-3p MIMAT0015766
          Rhesus monkey mml-miR-142-3p MIMAT0006200
          Xenopus laevis xla-miR-142-3p MIMAT0046455
          Xenopus tropicalis xtr-miR-142-3p MIMAT0003603
          Zebrafish dre-miR-142a-3p MIMAT0003160

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-142
          Stem-loop Accession # MI0000458
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000434
          miRBase ID: hsa-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-142a
          Stem-loop Accession # MI0000167
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000155
          miRBase ID: mmu-miR-142a-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-142
          Stem-loop Accession # MI0000915
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000848
          miRBase ID: rno-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-142-1
          Stem-loop Accession # MI0029746
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAAACAGCACUGCAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038163
          miRBase ID: ami-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Stem-loop ID ami-mir-142-2
          Stem-loop Accession # MI0029747
          Stem-loop Sequence
          CGGUGACAGUCUCCCCCCCCCAUAAAGUAGCGAGCACGACUCCGCCCCCGCGCCCGUGUAGUGUUUCCUACUUUAUGGAGGGGGUGGCUGGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038163
          miRBase ID: ami-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Artibeus jamaicensis

          Stem-loop ID aja-mir-142
          Stem-loop Accession # MI0024358
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031136
          miRBase ID: aja-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-142
          Stem-loop Accession # MI0028917
          Stem-loop Sequence
          UGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035200
          miRBase ID: efu-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-142
          Stem-loop Accession # MI0012771
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013023
          miRBase ID: eca-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-142
          Stem-loop Accession # MI0005301
          Stem-loop Sequence
          GCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCAAUGUAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004112
          miRBase ID: mdo-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-142
          Stem-loop Accession # MI0031355
          Stem-loop Sequence
          UAUGGACAGUGGACUCAUCCAUAAAGUAGAAAGCACUACUAAACGCUUGUGUCCAAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036722
          miRBase ID: oha-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-142
          Stem-loop Accession # MI0008543
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008037
          miRBase ID: ptr-miR-142
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-142
          Stem-loop Accession # MI0014833
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015766
          miRBase ID: ppy-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-142
          Stem-loop Accession # MI0007631
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006200
          miRBase ID: mml-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-142-2
          Stem-loop Accession # MI0038253
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAGACAGCACUGAACGCUGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046455
          miRBase ID: xla-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-142-1
          Stem-loop Accession # MI0004842
          Stem-loop Sequence
          CAGUGCAGCCACCCAUAAAGUAGAAAGCACUACUAGACAGGACUGAACGCUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003603
          miRBase ID: xtr-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Stem-loop ID xtr-mir-142-2
          Stem-loop Accession # MI0004843
          Stem-loop Sequence
          CAGUGCAGCCACCCAUAAAGUAGAAAGCACUACUAGACAGGACUGAACGCUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003603
          miRBase ID: xtr-miR-142-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-142a
          Stem-loop Accession # MI0002005
          Stem-loop Sequence
          CGUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACCCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003160
          miRBase ID: dre-miR-142a-3p
          Mature miRNA Sequence: UGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303162_pri
          TaqMan™ Pri-miRNA Assay : Rn03465064_pri
          TaqMan™ Pri-miRNA Assay : Mm03306331_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10398
          Ambion® Pre-miR™ miRNA Precursor : PM10398
          TaqMan™ MicroRNA Assay : 000464
          mirVana® miRNA inhibitor : MH10398
          mirVana® miRNA mimic : MC10398
          TaqMan™ Advanced miRNA Assay : mmu480934_mir
          TaqMan™ Advanced miRNA Assay : rno480934_mir
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel


          miRBase Alias:

          Mouse: mmu-miR-142-3p (v20)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline