Search
Search

| Assay Name: | mmu-miR-199a-3p | ||
|---|---|---|---|
| Stem-loop Accession Number: | MI0000242 | ||
| miRBase Version: | v22.1 | ||
| Chromosome Location: | Chr.19: 10817426 - 10817496 [-] on Build GRCh38 | ||
| Mature miRNA Sequence: | ACAGUAGUCUGCACAUUGGUUA | ||
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Bovine, Chrysemys picta, Dasypus novemcinctus, Gadus morhua, Guinea pig, Horse, Ophiophagus hannah, Oryzias latipes, Pan troglodytes, Paralichthys olivaceus, Pig, Rabbit, Rhesus monkey | ||
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | ACAGUAGUCUGCACAUUGGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-199a-3p | MIMAT0000232 |
| Human | hsa-miR-199b-3p | MIMAT0004563 |
| Mouse | mmu-miR-199a-3p | MIMAT0000230 |
| Mouse | mmu-miR-199b-3p | MIMAT0004667 |
| Rat | rno-miR-199a-3p | MIMAT0004738 |
| Alligator mississippiensis | ami-miR-199-3p | MIMAT0038217 |
| View More Rows | ||
| Bovine | bta-miR-199a-3p | MIMAT0003746 |
| Chrysemys picta | cpi-miR-199-3p | MIMAT0037819 |
| Dasypus novemcinctus | dno-miR-199-3p | MIMAT0047738 |
| Gadus morhua | gmo-miR-199-3p | MIMAT0044190 |
| Guinea pig | cpo-miR-199-3p | MIMAT0047087 |
| Horse | eca-miR-199a-3p | MIMAT0012961 |
| Horse | eca-miR-199b-3p | MIMAT0013781 |
| Ophiophagus hannah | oha-miR-199a-3p | MIMAT0036790 |
| Oryzias latipes | ola-miR-199a-3p | MIMAT0022539 |
| Pan troglodytes | ptr-miR-199a-3p | MIMAT0009192 |
| Pan troglodytes | ptr-miR-199b | MIMAT0008062 |
| Paralichthys olivaceus | pol-miR-199a-3p | MIMAT0025441 |
| Pig | ssc-miR-199a-3p | MIMAT0013875 |
| Rabbit | ocu-miR-199a-3p | MIMAT0048320 |
| Rhesus monkey | mml-miR-199a-3p | MIMAT0006232 |
| Stem-loop ID | hsa-mir-199a-1 |
| Stem-loop Accession # | MI0000242 |
| Stem-loop Sequence | GCCAACCCAGUGUUCAGACUACCUGUUCAGGAGGCUCUCAAUGUGUACAGUAGUCUGCACAUUGGUUAGGC |
| Chromosome Location | Chr. 19 - 10817426 - 10817496 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000232
miRBase ID: hsa-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 19 - 10817426 - 10817496 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-199a-2 |
| Stem-loop Accession # | MI0000281 |
| Stem-loop Sequence | AGGAAGCUUCUGGAGAUCCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGAGAGCA |
| Chromosome Location | Chr. 1 - 172144535 - 172144644 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000232
miRBase ID: hsa-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 19 - 10817426 - 10817496 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-199b |
| Stem-loop Accession # | MI0000282 |
| Stem-loop Sequence | CCAGAGGACACCUCCACUCCGUCUACCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGGGCUGGGUUAGACCCUCGG |
| Chromosome Location | Chr. 9 - 128244721 - 128244830 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004563
miRBase ID: hsa-miR-199b-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 9 - 128244721 - 128244830 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-199a-1 |
| Stem-loop Accession # | MI0000241 |
| Stem-loop Sequence | GCCAUCCCAGUGUUCAGACUACCUGUUCAGGAGGCUGGGACAUGUACAGUAGUCUGCACAUUGGUUAGGC |
| Chromosome Location | Chr. 9 - 21496495 - 21496564 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000230
miRBase ID: mmu-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 1 - 162217814 - 162217923 [+] on Build GRCm38 |
| Stem-loop ID | mmu-mir-199a-2 |
| Stem-loop Accession # | MI0000713 |
| Stem-loop Sequence | UGGAAGCUUCAGGAGAUCCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGCCAGCA |
| Chromosome Location | Chr. 1 - 162217814 - 162217923 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000230
miRBase ID: mmu-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 1 - 162217814 - 162217923 [+] on Build GRCm38 |
| Stem-loop ID | mmu-mir-199b |
| Stem-loop Accession # | MI0000714 |
| Stem-loop Sequence | CCAGAGGAUACCUCCACUCCGUCUACCCAGUGUUUAGACUACCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGGGCUGGGUUAGACCCUCGG |
| Chromosome Location | Chr. 2 - 32318460 - 32318569 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004667
miRBase ID: mmu-miR-199b-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 2 - 32318460 - 32318569 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-199a |
| Stem-loop Accession # | MI0000941 |
| Stem-loop Sequence | UGGAAGCUUCUGGAGAUCCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGCCAGCA |
| Chromosome Location | Chr. 13 - 80125487 - 80125596 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004738
miRBase ID: rno-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: Chr. 13 - 80125487 - 80125596 [+] on Build Rnor_6.0 |
| Stem-loop ID | ami-mir-199-1 |
| Stem-loop Accession # | MI0029782 |
| Stem-loop Sequence | AGUCCAGCCCAGCCUGCCCAGUGUUCAGACUACCUGUUCAGGAGCAUAUGAAGUUGUACAGUAGUCUGCACAUUGGUUAGAUCGGGCUUGGCAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038217
miRBase ID: ami-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ami-mir-199-2 |
| Stem-loop Accession # | MI0029783 |
| Stem-loop Sequence | CCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCUGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038217
miRBase ID: ami-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ami-mir-199-3 |
| Stem-loop Accession # | MI0029784 |
| Stem-loop Sequence | UCCACUCCGUCUGCCCAGUGUUCAGACUACCUGUUCAGGACUACGAGAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGUGCUGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038217
miRBase ID: ami-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | bta-mir-199a-1 |
| Stem-loop Accession # | MI0004758 |
| Stem-loop Sequence | UGGAAGCUUCUGGAGAUCCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003746
miRBase ID: bta-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | bta-mir-199a-2 |
| Stem-loop Accession # | MI0009770 |
| Stem-loop Sequence | GCCAACCCAGUGUUCAGACUACCUGUUCAGGGGGCUCUGAAUGUGUACAGUAGUCUGCACAUUGGUUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003746
miRBase ID: bta-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpi-mir-199-1 |
| Stem-loop Accession # | MI0029521 |
| Stem-loop Sequence | CCAGCCCAGCCUGCCCAGUGUUCAGACUACCUGUUCAGGAGGCUAUCAAGGUGUACAGUAGUCUGCACAUUGGUUAGAUUGGGUUUGGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037819
miRBase ID: cpi-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpi-mir-199-2 |
| Stem-loop Accession # | MI0029522 |
| Stem-loop Sequence | GAUCCUGCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCUGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCAAGGGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037819
miRBase ID: cpi-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpi-mir-199-3 |
| Stem-loop Accession # | MI0029523 |
| Stem-loop Sequence | GCUCCACUCCGUCUGCCCAGUGUUCAGACUACCUGUUCAGGACUACGAGAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGUGCUGGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037819
miRBase ID: cpi-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | dno-mir-199-1 |
| Stem-loop Accession # | MI0039032 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047738
miRBase ID: dno-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | dno-mir-199-2 |
| Stem-loop Accession # | MI0039033 |
| Stem-loop Sequence | CCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047738
miRBase ID: dno-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | gmo-mir-199-1 |
| Stem-loop Accession # | MI0036125 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGCUCUUACUGGUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044190
miRBase ID: gmo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | gmo-mir-199-2 |
| Stem-loop Accession # | MI0036108 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAUCAGGCUGCGUCUGAACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044190
miRBase ID: gmo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | gmo-mir-199-3 |
| Stem-loop Accession # | MI0036200 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGAAGUAGUGGUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044190
miRBase ID: gmo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-199-1 |
| Stem-loop Accession # | MI0038689 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGCGCCUCUGCAUGUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047087
miRBase ID: cpo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-199-2 |
| Stem-loop Accession # | MI0038690 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047087
miRBase ID: cpo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-199-3 |
| Stem-loop Accession # | MI0038691 |
| Stem-loop Sequence | CCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047087
miRBase ID: cpo-miR-199-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | eca-mir-199a |
| Stem-loop Accession # | MI0012712 |
| Stem-loop Sequence | GCUCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012961
miRBase ID: eca-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | eca-mir-199b |
| Stem-loop Accession # | MI0012922 |
| Stem-loop Sequence | GUCUACCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013781
miRBase ID: eca-miR-199b-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | oha-mir-199a |
| Stem-loop Accession # | MI0031392 |
| Stem-loop Sequence | UCUGCCCAGUGUUCGGACUACCUGUUCAGGACUACGAGAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036790
miRBase ID: oha-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ola-mir-199a-1 |
| Stem-loop Accession # | MI0019422 |
| Stem-loop Sequence | AGGCAUCCUGCUCCGUCAUCCCAGUGUUCAGACUACCUGUUCAGAAUCUUCCUGGUGUACAGUAGUCUGCACAUUGGUUAGACUGUGCAUGGAAGUAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022539
miRBase ID: ola-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ola-mir-199a-2 |
| Stem-loop Accession # | MI0019455 |
| Stem-loop Sequence | UCUCUAUCCCCCACCCCGCCUGCCCAGUGUUCAGACUACCUGUUCCUCCAGCUAUAACUGAACAGUAGUCUGCACAUUGGUUAGGCUAGACUGGGAGCCACACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022539
miRBase ID: ola-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ola-mir-199a-3 |
| Stem-loop Accession # | MI0019485 |
| Stem-loop Sequence | UACCCCUCCCCCCAUCCCGCCUGCCCAGUGUUCAGACUACCUGUUCAGGAAGUAGUGGUUGUACAGUAGUCUGCACAUUGGUUAGGCUGGUUAGGGAAGUGCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022539
miRBase ID: ola-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ola-mir-199a-4 |
| Stem-loop Accession # | MI0019518 |
| Stem-loop Sequence | AUUCCGGCUCCGUCCACCCAGUGUUCAGACUACCUGUUCAUUGUCAUACUGGUGUACAGUAGUCUGCACAUUGGUUAGACUGGGCCGGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022539
miRBase ID: ola-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-199a-1 |
| Stem-loop Accession # | MI0008571 |
| Stem-loop Sequence | GCCAACCCAGUGUUCAGACUACCUGUUCAGGAGGCUCUCAAUGUGUACAGUAGUCUGCACAUUGGUUAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009192
miRBase ID: ptr-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-199b |
| Stem-loop Accession # | MI0008572 |
| Stem-loop Sequence | CCAGAGGACACCUCCACUCCGUCUACCCAGUGUUUAGACUAUCUGUUCAGGACUCCCAAAUUGUACAGUAGUCUGCACAUUGGUUAGGCUGGGCUGGGUUAGACCCUCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008062
miRBase ID: ptr-miR-199b Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | pol-mir-199a-1 |
| Stem-loop Accession # | MI0022203 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAUUGUCAUACUGGUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0025441
miRBase ID: pol-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | pol-mir-199a-2 |
| Stem-loop Accession # | MI0022204 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGAAGUAGUGGUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0025441
miRBase ID: pol-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ssc-mir-199a-1 |
| Stem-loop Accession # | MI0016621 |
| Stem-loop Sequence | UCCGUCGCCCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUAGACUGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013875
miRBase ID: ssc-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ssc-mir-199a-2 |
| Stem-loop Accession # | MI0013093 |
| Stem-loop Sequence | CCCCGCCAACCCAGUGUUCAGACUACCUGUUCAGGGGGCUCUGAAUGUGUACAGUAGUCUGCACAUUGGUUAGGCUGGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013875
miRBase ID: ssc-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ocu-mir-199a-1 |
| Stem-loop Accession # | MI0039338 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGGGACUCCAGAUGUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048320
miRBase ID: ocu-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | ocu-mir-199a-2 |
| Stem-loop Accession # | MI0039339 |
| Stem-loop Sequence | CCCAGUGUUCAGACUACCUGUUCAGGACAAUGCCGUUGUACAGUAGUCUGCACAUUGGUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048320
miRBase ID: ocu-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |
| Stem-loop ID | mml-mir-199a-2 |
| Stem-loop Accession # | MI0007662 |
| Stem-loop Sequence | GCCAACCCAGUGUUCAGACUACCUGUUCAGGAGGCUCUCAACGUGUACAGUAGUCUGCACAUUGGUUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006232
miRBase ID: mml-miR-199a-3p Mature miRNA Sequence: ACAGUAGUCUGCACAUUGGUUA Chromosome Location: NA |