Search
Search

| Assay Name: | hsa-miR-199a-3p |
|---|---|
| miRBase Accession Number: | MI0000242 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAGUAGUCUGCACAUUGGUUA |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Bovine, Chrysemys picta, Dasypus novemcinctus, Gadus morhua, Guinea pig, Horse, Ophiophagus hannah, Oryzias latipes, Pan troglodytes, Paralichthys olivaceus, Pig, Rabbit, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | ACAGUAGUCUGCACAUUGGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-199a-3p | MIMAT0000232 |
| Human | hsa-miR-199b-3p | MIMAT0004563 |
| Mouse | mmu-miR-199a-3p | MIMAT0000230 |
| Mouse | mmu-miR-199b-3p | MIMAT0004667 |
| Rat | rno-miR-199a-3p | MIMAT0004738 |
| Alligator mississippiensis | ami-miR-199-3p | MIMAT0038217 |
| View More Rows | ||
| Bovine | bta-miR-199a-3p | MIMAT0003746 |
| Chrysemys picta | cpi-miR-199-3p | MIMAT0037819 |
| Dasypus novemcinctus | dno-miR-199-3p | MIMAT0047738 |
| Gadus morhua | gmo-miR-199-3p | MIMAT0044190 |
| Guinea pig | cpo-miR-199-3p | MIMAT0047087 |
| Horse | eca-miR-199a-3p | MIMAT0012961 |
| Horse | eca-miR-199b-3p | MIMAT0013781 |
| Ophiophagus hannah | oha-miR-199a-3p | MIMAT0036790 |
| Oryzias latipes | ola-miR-199a-3p | MIMAT0022539 |
| Pan troglodytes | ptr-miR-199a-3p | MIMAT0009192 |
| Pan troglodytes | ptr-miR-199b | MIMAT0008062 |
| Paralichthys olivaceus | pol-miR-199a-3p | MIMAT0025441 |
| Pig | ssc-miR-199a-3p | MIMAT0013875 |
| Rabbit | ocu-miR-199a-3p | MIMAT0048320 |
| Rhesus monkey | mml-miR-199a-3p | MIMAT0006232 |