Search
Search

| Assay Name: | mmu-miR-196a-1-3p |
|---|---|
| Stem-loop Accession Number: | MI0000552 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAACGACAUCAAACCACCUGAU |
| Species: | Mouse |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | CAACGACAUCAAACCACCUGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-196a-1-3p | MIMAT0017013 |