Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482715_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-24-1-5p
          Stem-loop Accession Number: MI0000231
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 63301208 - 63301275 [+] on Build GRCm38
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Species: Mouse, Bovine, Chicken, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Macaca nemestrina, Ophiophagus hannah, Pan paniscus, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482715_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000041, mir-24

          Mature miRNA Sequence:

          GUGCCUACUGAGCUGAUAUCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-24-1-5p MIMAT0000218
          Bovine bta-miR-24 MIMAT0009250
          Chicken gga-miR-24-5p MIMAT0026548
          Cricetulus griseus cgr-miR-24 MIMAT0041158
          Dasypus novemcinctus dno-miR-24a-5p MIMAT0047532
          Guinea pig cpo-miR-24-5p MIMAT0046880
          View More Rows
          Macaca nemestrina mne-miR-24-5p MIMAT0002340
          Ophiophagus hannah oha-miR-24-1-5p MIMAT0036858
          Pan paniscus ppa-miR-24-5p MIMAT0002342
          Pig ssc-miR-24-2-5p MIMAT0026468
          Pongo pygmaeus ppy-miR-24-5p MIMAT0002338
          Rabbit ocu-miR-24-5p MIMAT0048130
          Rhesus monkey mml-miR-24-5p MIMAT0002336
          Taeniopygia guttata tgu-miR-24-5p MIMAT0026994

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-24-1
          Stem-loop Accession # MI0000231
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000218
          miRBase ID: mmu-miR-24-1-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-24-1
          Stem-loop Accession # MI0009761
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009250
          miRBase ID: bta-miR-24
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-24
          Stem-loop Accession # MI0001275
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUACAUACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026548
          miRBase ID: gga-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-24-1
          Stem-loop Accession # MI0033087
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041158
          miRBase ID: cgr-miR-24
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-24a
          Stem-loop Accession # MI0038921
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047532
          miRBase ID: dno-miR-24a-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-24-1
          Stem-loop Accession # MI0038577
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCUUGUACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046880
          miRBase ID: cpo-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-24-1
          Stem-loop Accession # MI0002639
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002340
          miRBase ID: mne-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-24-1
          Stem-loop Accession # MI0031432
          Stem-loop Sequence
          CGUCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUAUAUACUGGCUCAGUUCAGCAGGAACAGGAGUCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036858
          miRBase ID: oha-miR-24-1-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-24-1
          Stem-loop Accession # MI0002640
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002342
          miRBase ID: ppa-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-24-2
          Stem-loop Accession # MI0013111
          Stem-loop Sequence
          GACCCGCCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAGUCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026468
          miRBase ID: ssc-miR-24-2-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-24-1
          Stem-loop Accession # MI0002638
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002338
          miRBase ID: ppy-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-24-1
          Stem-loop Accession # MI0039236
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCGUUUCACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048130
          miRBase ID: ocu-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-24-1
          Stem-loop Accession # MI0002637
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002336
          miRBase ID: mml-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-24
          Stem-loop Accession # MI0013753
          Stem-loop Sequence
          CCAGUCGGAAACAACUUACAGCUGCUGCUGUUGUUCCAGGCUGUUGAUGGACCCGUCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAGUCGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026994
          miRBase ID: tgu-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306468_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10073
          Ambion® Pre-miR™ miRNA Precursor : PM10073
          TaqMan™ MicroRNA Assay : 000488
          mirVana® miRNA inhibitor : MH10073
          mirVana® miRNA mimic : MC10073

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline