Search
Search

| Assay Name: | rno-miR-200a-3p |
|---|---|
| Stem-loop Accession Number: | MI0000737 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAACACUGUCUGGUAACGAUGU |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chicken, Fugu rubripes, Gadus morhua, Gorilla gorilla, Horse, Monodelphis domestica, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | UAACACUGUCUGGUAACGAUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-200a-3p | MIMAT0000682 |
| Mouse | mmu-miR-200a-3p | MIMAT0000519 |
| Rat | rno-miR-200a-3p | MIMAT0000874 |
| Alligator mississippiensis | ami-miR-200a-3p | MIMAT0038219 |
| Anolis carolinensis | aca-miR-200a-3p | MIMAT0021845 |
| Chicken | gga-miR-200a-3p | MIMAT0001171 |
| View More Rows | ||
| Fugu rubripes | fru-miR-200a | MIMAT0002981 |
| Gadus morhua | gmo-miR-200a-3p | MIMAT0044106 |
| Gorilla gorilla | ggo-miR-200a | MIMAT0024300 |
| Horse | eca-miR-200a | MIMAT0012909 |
| Monodelphis domestica | mdo-miR-200a-3p | MIMAT0004158 |
| Pan troglodytes | ptr-miR-200a | MIMAT0008063 |
| Pongo pygmaeus | ppy-miR-200a | MIMAT0015796 |
| Rhesus monkey | mml-miR-200a-3p | MIMAT0006233 |
| Tetraodon nigroviridis | tni-miR-200a | MIMAT0002982 |
| Xenopus tropicalis | xtr-miR-200a | MIMAT0003693 |
| Zebrafish | dre-miR-200a-3p | MIMAT0001861 |