Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10991
          Assay Name: hsa-miR-200a-3p
          miRBase Accession Number: MI0000737
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 1167863 - 1167952 [+] on Build GRCh38
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chicken, Fugu rubripes, Gadus morhua, Gorilla gorilla, Horse, Monodelphis domestica, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC10991

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAACACUGUCUGGUAACGAUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200a-3p MIMAT0000682
          Mouse mmu-miR-200a-3p MIMAT0000519
          Rat rno-miR-200a-3p MIMAT0000874
          Alligator mississippiensis ami-miR-200a-3p MIMAT0038219
          Anolis carolinensis aca-miR-200a-3p MIMAT0021845
          Chicken gga-miR-200a-3p MIMAT0001171
          View More Rows
          Fugu rubripes fru-miR-200a MIMAT0002981
          Gadus morhua gmo-miR-200a-3p MIMAT0044106
          Gorilla gorilla ggo-miR-200a MIMAT0024300
          Horse eca-miR-200a MIMAT0012909
          Monodelphis domestica mdo-miR-200a-3p MIMAT0004158
          Pan troglodytes ptr-miR-200a MIMAT0008063
          Pongo pygmaeus ppy-miR-200a MIMAT0015796
          Rhesus monkey mml-miR-200a-3p MIMAT0006233
          Tetraodon nigroviridis tni-miR-200a MIMAT0002982
          Xenopus tropicalis xtr-miR-200a MIMAT0003693
          Zebrafish dre-miR-200a-3p MIMAT0001861

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200a
          Stem-loop Accession # MI0000737
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCGC
          Chromosome Location Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000682
          miRBase ID: hsa-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200a
          Stem-loop Accession # MI0000554
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000519
          miRBase ID: mmu-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-200a
          Stem-loop Accession # MI0000943
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCA
          Chromosome Location Chr. 5 - 173489366 - 173489454 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000874
          miRBase ID: rno-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: Chr. 5 - 173489366 - 173489454 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-200a
          Stem-loop Accession # MI0029785
          Stem-loop Sequence
          UGGUCCUCUGUGGGCAUCUUACUAGACAGUGCUGGAUAUUUUGGAUCUACUCUAACACUGUCUGGUAACGAUGUUUAAAGGGUGAACCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038219
          miRBase ID: ami-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-200a
          Stem-loop Accession # MI0018802
          Stem-loop Sequence
          ACGAUGAUGGUCCUCUGUGGACAUCUUACUAGACAGUGCUGGAUUGUGUGCUCUGAUUCUAACACUGUCUGGUAACGAUGUUCAAAGGGUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021845
          miRBase ID: aca-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-200a
          Stem-loop Accession # MI0001249
          Stem-loop Sequence
          GGUCCUCUGUGGGCAUCUUACUAGACAGUGCUGGAUUUCUUGGAUCUAUUCUAACACUGUCUGGUAACGAUGUUUAAAGGGUGAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001171
          miRBase ID: gga-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-200a
          Stem-loop Accession # MI0003303
          Stem-loop Sequence
          UCUCAGGAUCCAUCUUACCCGACAGUGCUGGAUUGUACUACUGUUGUUCUAACACUGUCUGGUAACGAUGUUUUCUGGGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002981
          miRBase ID: fru-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-200a
          Stem-loop Accession # MI0036045
          Stem-loop Sequence
          CAUCUUACCUGGCAGUGCUUGAUUGUAUGACUGGUCUUCUAACACUGUCUGGUAACGAUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044106
          miRBase ID: gmo-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-200a
          Stem-loop Accession # MI0020848
          Stem-loop Sequence
          GGGACCCCACGUCCCUCCCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCGCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024300
          miRBase ID: ggo-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-200a
          Stem-loop Accession # MI0012665
          Stem-loop Sequence
          CUGGGCCCCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUCGGCUCGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012909
          miRBase ID: eca-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-200a
          Stem-loop Accession # MI0005346
          Stem-loop Sequence
          GGGCCUCUGUGGGCAUCUUACUAGACAGUGCUGGAUUUUUGGAUGUACUCUAACACUGUCUGGUAACGAUGUUUAAAGAGGGAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004158
          miRBase ID: mdo-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-200a
          Stem-loop Accession # MI0008573
          Stem-loop Sequence
          CGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008063
          miRBase ID: ptr-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-200a
          Stem-loop Accession # MI0014861
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015796
          miRBase ID: ppy-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-200a
          Stem-loop Accession # MI0007663
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006233
          miRBase ID: mml-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-200a
          Stem-loop Accession # MI0003304
          Stem-loop Sequence
          CAUCUUACCUGACAGUGCUGGAUUAUACUACUGUUGUUCUAACACUGUCUGGUAACGAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002982
          miRBase ID: tni-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-200a
          Stem-loop Accession # MI0004945
          Stem-loop Sequence
          UGGUCCUCUAUGGACAUCUUACUAGACAGUGCUGGAUUUAUUUUAUCUUUUCUAACACUGUCUGGUAACGAUGUUUAAAGAGUGAGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003693
          miRBase ID: xtr-miR-200a
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-200a
          Stem-loop Accession # MI0002037
          Stem-loop Sequence
          GGCACUUAGCAGCCAUCUUACCGGACAGUGCUGGACUGUAUAACUGUUUUCUAACACUGUCUGGUAACGAUGUUUGUUGGGUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001861
          miRBase ID: dre-miR-200a-3p
          Mature miRNA Sequence: UAACACUGUCUGGUAACGAUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306736_pri
          TaqMan™ Pri-miRNA Assay : Rn03464802_pri
          TaqMan™ Pri-miRNA Assay : Hs03303376_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10991
          Ambion® Pre-miR™ miRNA Precursor : PM10991
          TaqMan™ MicroRNA Assay : 000502
          mirVana® miRNA inhibitor : MH10991
          TaqMan™ Advanced miRNA Assay : rno478490_mir
          TaqMan™ Advanced miRNA Assay : mmu478490_mir
          TaqMan™ Advanced miRNA Assay : 478490_mir


          miRBase Alias:

          Human: hsa-miR-200a (v17)
          Mouse: mmu-miR-200a (v17)
          Rat: rno-miR-200a (v18)
          Aeromonas caviae Ae398: aca-miR-200a (v18)
          Danio rerio: dre-miR-200a (v20)
          Gallus gallus: gga-miR-200a (v19)
          Macaca mulatta: mml-miR-200a (v19)
          Monodelphis domestica: mdo-miR-200a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline