Search
Search

| Assay Name: | rno-miR-192-5p |
|---|---|
| Stem-loop Accession Number: | MI0000234 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGACCUAUGAAUUGACAGCC |
| Species: | Human, Mouse, Rat, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | CUGACCUAUGAAUUGACAGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-192-5p | MIMAT0000222 |
| Mouse | mmu-miR-192-5p | MIMAT0000517 |
| Rat | rno-miR-192-5p | MIMAT0000867 |
| Dog | cfa-miR-192 | MIMAT0006632 |
| Goat | chi-miR-192-5p | MIMAT0036024 |
| Gorilla gorilla | ggo-miR-192 | MIMAT0024119 |
| View More Rows | ||
| Guinea pig | cpo-miR-192-5p | MIMAT0047068 |
| Horse | eca-miR-192 | MIMAT0013049 |
| Nomascus leucogenys | nle-miR-192 | MIMAT0049384 |
| Ornithorhynchus anatinus | oan-miR-192-5p | MIMAT0007016 |
| Otolemur garnettii | oga-miR-192 | MIMAT0049704 |
| Pan paniscus | ppa-miR-192 | MIMAT0049059 |
| Pan troglodytes | ptr-miR-192 | MIMAT0008057 |
| Pig | ssc-miR-192 | MIMAT0013910 |
| Pongo pygmaeus | ppy-miR-192 | MIMAT0015786 |
| Rhesus monkey | mml-miR-192-5p | MIMAT0006224 |
| Tupaia chinensis | tch-miR-192-5p | MIMAT0036616 |