Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480882_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-195-5p
          Stem-loop Accession Number: MI0000489
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 7017615 - 7017701 [-] on Build GRCh38
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Species: Human, Mouse, Rat, Gorilla gorilla, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480882_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          UAGCAGCACAGAAAUAUUGGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-195-5p MIMAT0000461
          Mouse mmu-miR-195a-5p MIMAT0000225
          Rat rno-miR-195-5p MIMAT0000870
          Gorilla gorilla ggo-miR-195 MIMAT0002316
          Horse eca-miR-195 MIMAT0013027
          Pan paniscus ppa-miR-195 MIMAT0002317
          View More Rows
          Pan troglodytes ptr-miR-195 MIMAT0008060
          Pig ssc-miR-195 MIMAT0013928
          Pongo pygmaeus ppy-miR-195 MIMAT0015790
          Rhesus monkey mml-miR-195-5p MIMAT0006228

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-195
          Stem-loop Accession # MI0000489
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location Chr. 17 - 7017615 - 7017701 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000461
          miRBase ID: hsa-miR-195-5p
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: Chr. 17 - 7017615 - 7017701 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-195a
          Stem-loop Accession # MI0000237
          Stem-loop Sequence
          ACACCCAACUCUCCUGGCUCUAGCAGCACAGAAAUAUUGGCAUGGGGAAGUGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUGA
          Chromosome Location Chr. 11 - 70235042 - 70235135 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000225
          miRBase ID: mmu-miR-195a-5p
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: Chr. 11 - 70235042 - 70235135 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-195
          Stem-loop Accession # MI0000939
          Stem-loop Sequence
          AACUCUCCUGGCUCUAGCAGCACAGAAAUAUUGGCACGGGUAAGUGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000870
          miRBase ID: rno-miR-195-5p
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0

          Gorilla gorilla

          Stem-loop ID ggo-mir-195
          Stem-loop Accession # MI0002617
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002316
          miRBase ID: ggo-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-195
          Stem-loop Accession # MI0012774
          Stem-loop Sequence
          ACACCCAGCUCCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACUGGGAAGAGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013027
          miRBase ID: eca-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-195
          Stem-loop Accession # MI0002618
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002317
          miRBase ID: ppa-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-195
          Stem-loop Accession # MI0008568
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008060
          miRBase ID: ptr-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-195
          Stem-loop Accession # MI0013141
          Stem-loop Sequence
          CAGCCCCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACCGGGCGGAGAGUCUGCCAGUAUUGGCUGUGCUGCCCCAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013928
          miRBase ID: ssc-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-195
          Stem-loop Accession # MI0014857
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015790
          miRBase ID: ppy-miR-195
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-195
          Stem-loop Accession # MI0007658
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCAAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006228
          miRBase ID: mml-miR-195-5p
          Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306496_pri
          TaqMan™ Pri-miRNA Assay : Rn03465102_pri
          TaqMan™ Pri-miRNA Assay : Hs03303320_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10827
          Ambion® Pre-miR™ miRNA Precursor : PM10827
          TaqMan™ MicroRNA Assay : 000494
          mirVana® miRNA inhibitor : MH10827
          mirVana® miRNA mimic : MC10827
          TaqMan™ Advanced miRNA Assay : 477957_mir
          TaqMan™ Advanced miRNA Assay : mmu482953_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline