Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn03465102_pri
Assay Name:
rno-mir-195
Stem-loop ID:
rno-mir-195
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.10: 56845301 - 56845387 [+] on Build Rnor_6.0
Stem-loop Location:
Chr.10: 56845301 - 56845387 [+] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-195-5p
rno-miR-195-3p
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn03465102_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
68
Chromosome Location:
Chr. 10 - 56845755 - 56845755 [-] on 56845755 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-195-5p
MIMAT0000870
Rat
rno-miR-195-3p
MIMAT0017149
Stem-loop Details
Rat
Stem-loop ID
rno-mir-195
Stem-loop Accession #
MI0000939
Stem-loop Sequence
AACUCUCCUGGCUCUAGCAGCACAGAAAUAUUGGCACGGGUAAGUGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
Chromosome Location
Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0000870
miRBase ID: rno-miR-195-5p
Mature miRNA Sequence: UAGCAGCACAGAAAUAUUGGC
Chromosome Location: Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0
miRBase Accession #: MIMAT0017149
miRBase ID: rno-miR-195-3p
Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
Chromosome Location: Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0
Gene Family ID
MIPF0000006,
mir-15
Distance to Amplicon
331 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10827
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10827
TaqMan
™
MicroRNA Assay :
000494
mir
Vana
®
miRNA inhibitor :
MH10827
mir
Vana
®
miRNA mimic :
MC10827
TaqMan
™
Advanced miRNA Assay :
477957_mir
TaqMan
™
Advanced miRNA Assay :
mmu482953_mir
TaqMan
™
Advanced miRNA Assay :
rno480882_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19206
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19206
mir
Vana
®
miRNA inhibitor :
MH19206
mir
Vana
®
miRNA mimic :
MC19206
TaqMan
™
Advanced miRNA Assay :
rno480978_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline