Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480978_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-195-3p
          Stem-loop Accession Number: MI0000939
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 56845301 - 56845387 [+] on Build Rnor_6.0
          Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
          Species: Rat, Dasypus novemcinctus, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480978_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          CCAAUAUUGGCUGUGCUGCUCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-195-3p MIMAT0017149
          Dasypus novemcinctus dno-miR-195-3p MIMAT0047728
          Rabbit ocu-miR-195-3p MIMAT0048310
          Rhesus monkey mml-miR-195-3p MIMAT0026838

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-195
          Stem-loop Accession # MI0000939
          Stem-loop Sequence
          AACUCUCCUGGCUCUAGCAGCACAGAAAUAUUGGCACGGGUAAGUGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017149
          miRBase ID: rno-miR-195-3p
          Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
          Chromosome Location: Chr. 10 - 56845301 - 56845387 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-195
          Stem-loop Accession # MI0039027
          Stem-loop Sequence
          UAGCAGCACAGAAAUAUUGGCACUGGGAAGAAAGUCUGCCAAUAUUGGCUGUGCUGCUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047728
          miRBase ID: dno-miR-195-3p
          Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-195
          Stem-loop Accession # MI0039333
          Stem-loop Sequence
          UAGCAGCACAGAAAUAUUGGCACCGGGAGGCGAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048310
          miRBase ID: ocu-miR-195-3p
          Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-195
          Stem-loop Accession # MI0007658
          Stem-loop Sequence
          AGCUUCCCUGGCUCUAGCAGCACAGAAAUAUUGGCACAGGGAAGCAAGUCUGCCAAUAUUGGCUGUGCUGCUCCAGGCAGGGUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026838
          miRBase ID: mml-miR-195-3p
          Mature miRNA Sequence: CCAAUAUUGGCUGUGCUGCUCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465102_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19206
          Ambion® Pre-miR™ miRNA Precursor : PM19206
          mirVana® miRNA inhibitor : MH19206
          mirVana® miRNA mimic : MC19206

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline