Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480938_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-145-5p
          Stem-loop Accession Number: MI0000461
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 149430646 - 149430733 [+] on Build GRCh38
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Daubentonia madagascariensis, Dog, Gadus morhua, Goat, Guinea pig, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Papio hamadryas, Petromyzon marinus, Pteropus alecto, Python bivittatus, Rabbit, Salmo salar
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480938_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000079, mir-145

          Mature miRNA Sequence:

          GUCCAGUUUUCCCAGGAAUCCCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-145-5p MIMAT0000437
          Mouse mmu-miR-145a-5p MIMAT0000157
          Rat rno-miR-145-5p MIMAT0000851
          Bovine bta-miR-145 MIMAT0003542
          Callithrix jacchus cja-miR-145 MIMAT0039414
          Chrysemys picta cpi-miR-145-5p MIMAT0037772
          View More Rows
          Columba livia cli-miR-145-5p MIMAT0038533
          Daubentonia madagascariensis dma-miR-145 MIMAT0049256
          Dog cfa-miR-145 MIMAT0009863
          Gadus morhua gmo-miR-145-5p MIMAT0044334
          Goat chi-miR-145-5p MIMAT0035967
          Guinea pig cpo-miR-145-5p MIMAT0047014
          Horse eca-miR-145 MIMAT0013064
          Ictalurus punctatus ipu-miR-145 MIMAT0029422
          Microcebus murinus mmr-miR-145 MIMAT0049208
          Monodelphis domestica mdo-miR-145-5p MIMAT0004116
          Nomascus leucogenys nle-miR-145 MIMAT0049426
          Oreochromis niloticus oni-miR-145 MIMAT0043395
          Otolemur garnettii oga-miR-145 MIMAT0049668
          Pan paniscus ppa-miR-145 MIMAT0049057
          Papio hamadryas pha-miR-145 MIMAT0049513
          Petromyzon marinus pma-miR-145-5p MIMAT0019476
          Pteropus alecto pal-miR-145-5p MIMAT0040158
          Python bivittatus pbv-miR-145-5p MIMAT0038959
          Rabbit ocu-miR-145-5p MIMAT0048251
          Salmo salar ssa-miR-145-5p MIMAT0032369

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-145
          Stem-loop Accession # MI0000461
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000437
          miRBase ID: hsa-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-145a
          Stem-loop Accession # MI0000169
          Stem-loop Sequence
          CUCACGGUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG
          Chromosome Location Chr. 18 - 61647825 - 61647894 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000157
          miRBase ID: mmu-miR-145a-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: Chr. 18 - 61647825 - 61647894 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-145
          Stem-loop Accession # MI0000918
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGCU
          Chromosome Location Chr. 18 - 56969907 - 56969994 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000851
          miRBase ID: rno-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: Chr. 18 - 56969907 - 56969994 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-145
          Stem-loop Accession # MI0004756
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003542
          miRBase ID: bta-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-145
          Stem-loop Accession # MI0031934
          Stem-loop Sequence
          ACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039414
          miRBase ID: cja-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-145
          Stem-loop Accession # MI0029488
          Stem-loop Sequence
          GUGUCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCACUAUGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCAUGGCUCAGCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037772
          miRBase ID: cpi-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-145
          Stem-loop Accession # MI0029979
          Stem-loop Sequence
          CCGUGCCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCGCUACGUUGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038533
          miRBase ID: cli-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-145
          Stem-loop Accession # MI0039984
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049256
          miRBase ID: dma-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-145
          Stem-loop Accession # MI0010359
          Stem-loop Sequence
          UGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009863
          miRBase ID: cfa-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-145
          Stem-loop Accession # MI0036228
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGUUGAACAUGAAAGCGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044334
          miRBase ID: gmo-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-145
          Stem-loop Accession # MI0030631
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035967
          miRBase ID: chi-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-145
          Stem-loop Accession # MI0038652
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047014
          miRBase ID: cpo-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-145
          Stem-loop Accession # MI0012808
          Stem-loop Sequence
          CUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013064
          miRBase ID: eca-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-145
          Stem-loop Accession # MI0024511
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCGAAAGGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029422
          miRBase ID: ipu-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-145
          Stem-loop Accession # MI0039931
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049208
          miRBase ID: mmr-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-145
          Stem-loop Accession # MI0005305
          Stem-loop Sequence
          CUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004116
          miRBase ID: mdo-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-145
          Stem-loop Accession # MI0040168
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049426
          miRBase ID: nle-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-145
          Stem-loop Accession # MI0035432
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGACCUAUCAGAAAGGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043395
          miRBase ID: oni-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-145
          Stem-loop Accession # MI0040451
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049668
          miRBase ID: oga-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-145
          Stem-loop Accession # MI0039765
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049057
          miRBase ID: ppa-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-145
          Stem-loop Accession # MI0040278
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAAAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049513
          miRBase ID: pha-miR-145
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-145
          Stem-loop Accession # MI0017080
          Stem-loop Sequence
          UGCGUGGGCAUGGUCCAGUUUUCCCAGGAAUCCCUGUGCCCUUUUCCCGGGAUUCCAGGAAUAACUGUCCAAGCACACGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019476
          miRBase ID: pma-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-145
          Stem-loop Accession # MI0032524
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040158
          miRBase ID: pal-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-145
          Stem-loop Accession # MI0030243
          Stem-loop Sequence
          ACUAUGUCCUCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUCAUGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038959
          miRBase ID: pbv-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-145
          Stem-loop Accession # MI0039303
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048251
          miRBase ID: ocu-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-145-1
          Stem-loop Accession # MI0026505
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCAACAACGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032369
          miRBase ID: ssa-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-145-2
          Stem-loop Accession # MI0026506
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCACAAGGGGGAUUCCUGGAAAUACUGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032369
          miRBase ID: ssa-miR-145-5p
          Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303169_pri
          TaqMan™ Pri-miRNA Assay : Rn03466035_pri
          TaqMan™ Pri-miRNA Assay : Mm03306347_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11480
          Ambion® Pre-miR™ miRNA Precursor : PM11480
          TaqMan™ MicroRNA Assay : 002278
          mirVana® miRNA inhibitor : MH11480
          mirVana® miRNA mimic : MC11480
          TaqMan™ Advanced miRNA Assay : 477916_mir
          TaqMan™ Advanced miRNA Assay : mmu480938_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline