Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481005_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-221-3p
          Stem-loop Accession Number: MI0000298
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 45746157 - 45746266 [-] on Build GRCh38
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Chicken, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Fugu rubripes, Gorilla gorilla, Guinea pig, Horse, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Oryzias latipes, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pundamilia nyererei, Rabbit, Rhesus monkey, Saimiri boliviensis, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481005_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000051, mir-221

          Mature miRNA Sequence:

          AGCUACAUUGUCUGCUGGGUUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-221-3p MIMAT0000278
          Mouse mmu-miR-221-3p MIMAT0000669
          Rat rno-miR-221-3p MIMAT0000890
          Alligator mississippiensis ami-miR-221-3p MIMAT0038253
          Astatotilapia burtoni abu-miR-221 MIMAT0042073
          Chicken gga-miR-221-3p MIMAT0001108
          View More Rows
          Cricetulus griseus cgr-miR-221-3p MIMAT0023859
          Dasypus novemcinctus dno-miR-221-3p MIMAT0047776
          Daubentonia madagascariensis dma-miR-221 MIMAT0049278
          Fugu rubripes fru-miR-221 MIMAT0002906
          Gorilla gorilla ggo-miR-221 MIMAT0002584
          Guinea pig cpo-miR-221-3p MIMAT0047131
          Horse eca-miR-221 MIMAT0013203
          Metriaclima zebra mze-miR-221 MIMAT0042316
          Microcebus murinus mmr-miR-221 MIMAT0049144
          Monodelphis domestica mdo-miR-221-3p MIMAT0004160
          Neolamprologus brichardi nbr-miR-221 MIMAT0042495
          Nomascus leucogenys nle-miR-221 MIMAT0049404
          Ophiophagus hannah oha-miR-221-3p MIMAT0036847
          Oreochromis niloticus oni-miR-221 MIMAT0042675
          Oryzias latipes ola-miR-221 MIMAT0022530
          Otolemur garnettii oga-miR-221 MIMAT0049658
          Pan paniscus ppa-miR-221 MIMAT0002586
          Pan troglodytes ptr-miR-221 MIMAT0008076
          Papio hamadryas pha-miR-221 MIMAT0049559
          Pongo pygmaeus ppy-miR-221 MIMAT0002585
          Pundamilia nyererei pny-miR-221 MIMAT0042889
          Rabbit ocu-miR-221-3p MIMAT0048355
          Rhesus monkey mml-miR-221-3p MIMAT0002583
          Saimiri boliviensis sbo-miR-221 MIMAT0049481
          Taeniopygia guttata tgu-miR-221-3p MIMAT0014512
          Tetraodon nigroviridis tni-miR-221 MIMAT0002907
          Xenopus tropicalis xtr-miR-221 MIMAT0003700
          Zebrafish dre-miR-221-3p MIMAT0001288

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-221
          Stem-loop Accession # MI0000298
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location Chr. X - 45746157 - 45746266 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000278
          miRBase ID: hsa-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: Chr. X - 45746157 - 45746266 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-221
          Stem-loop Accession # MI0000709
          Stem-loop Sequence
          AUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAA
          Chromosome Location Chr. X - 19146294 - 19146388 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000669
          miRBase ID: mmu-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: Chr. X - 19146294 - 19146388 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-221
          Stem-loop Accession # MI0000961
          Stem-loop Sequence
          UGAAUAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAGCAUUUCUC
          Chromosome Location Chr. X - 3684480 - 3684588 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000890
          miRBase ID: rno-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: Chr. X - 3684480 - 3684588 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-221
          Stem-loop Accession # MI0029806
          Stem-loop Sequence
          UCCAGGUUUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUAUUUCUUAAGCAACAGCUACAUUGUCUGCUGGGUUUCAAGCUGCCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038253
          miRBase ID: ami-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-221-1
          Stem-loop Accession # MI0033768
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUGGUUCAAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042073
          miRBase ID: abu-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Stem-loop ID abu-mir-221-2
          Stem-loop Accession # MI0033779
          Stem-loop Sequence
          ACCUGGCAUACAGUGUAGGAAUCUGUGUGUGUCAGUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042073
          miRBase ID: abu-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-221
          Stem-loop Accession # MI0001178
          Stem-loop Sequence
          GACUGUCCAGGUUUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAAGCAACAGCUACAUUGUCUGCUGGGUUUCCAGCUGCCUGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001108
          miRBase ID: gga-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-221
          Stem-loop Accession # MI0020464
          Stem-loop Sequence
          GGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUAUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023859
          miRBase ID: cgr-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-221
          Stem-loop Accession # MI0039052
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUGGACAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047776
          miRBase ID: dno-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-221
          Stem-loop Accession # MI0040008
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049278
          miRBase ID: dma-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-221
          Stem-loop Accession # MI0003221
          Stem-loop Sequence
          CCUGAACCUGGCAUACAAUGUAGAUUUCUGUGUGCUUGGAUUCUACAGCUACAUUGUCUGCUGGGUUUCCGGAUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002906
          miRBase ID: fru-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-221
          Stem-loop Accession # MI0002893
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002584
          miRBase ID: ggo-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-221
          Stem-loop Accession # MI0038713
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUGAACAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047131
          miRBase ID: cpo-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-221
          Stem-loop Accession # MI0012951
          Stem-loop Sequence
          CCAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUCUUGGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013203
          miRBase ID: eca-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-221-1
          Stem-loop Accession # MI0034081
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUGGUUCAAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042316
          miRBase ID: mze-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Stem-loop ID mze-mir-221-2
          Stem-loop Accession # MI0034085
          Stem-loop Sequence
          ACCUGGCAUACAGUGUAGGAAUCUGUGUGUGUCAGUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042316
          miRBase ID: mze-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-221
          Stem-loop Accession # MI0039858
          Stem-loop Sequence
          ACCUGGCACACAAUGUAGAUUUCUGUGUUUGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049144
          miRBase ID: mmr-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-221
          Stem-loop Accession # MI0005348
          Stem-loop Sequence
          GCAUGAACCUGGCAUACAAUGUAGAAUUCUGUGUUUAUUAAGUAACAGCUACAUUGUCUGCUGGGUUUCAGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004160
          miRBase ID: mdo-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-221-1
          Stem-loop Accession # MI0034327
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUGGUUCAAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042495
          miRBase ID: nbr-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-221-2
          Stem-loop Accession # MI0034335
          Stem-loop Sequence
          ACCUGGCAUACAGUGUAGGAAUCUGUGUGUGUCAGUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042495
          miRBase ID: nbr-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-221
          Stem-loop Accession # MI0040144
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUCAUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049404
          miRBase ID: nle-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-221
          Stem-loop Accession # MI0031426
          Stem-loop Sequence
          GAUGAUCCAGCUUUUGGGCCUUCACCUGGCAUACAAUGUAGAAUACUGUGUUUGUUAAGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUGUCUGGAAGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036847
          miRBase ID: oha-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-221-1
          Stem-loop Accession # MI0034576
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUGGUUCUAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042675
          miRBase ID: oni-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Stem-loop ID oni-mir-221-2
          Stem-loop Accession # MI0034580
          Stem-loop Sequence
          ACCUGGCAUACAGUGUAGGAAUCUGUGUGUGUCAGUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042675
          miRBase ID: oni-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-221
          Stem-loop Accession # MI0019415
          Stem-loop Sequence
          CCGUCCAUGAGGAGUUCUGGACCUGGCAUACAGUGUAGUAUUCUGUGUGCGUCCGUCUACAGCUACAUUGUCUGCUGGGUUUCAGGCCUCCACUCAAGUCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022530
          miRBase ID: ola-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-221
          Stem-loop Accession # MI0040441
          Stem-loop Sequence
          ACCUGGCACACAAUGUAGAUUUCUGUGUUAGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049658
          miRBase ID: oga-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-221
          Stem-loop Accession # MI0002895
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002586
          miRBase ID: ppa-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-221
          Stem-loop Accession # MI0008587
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008076
          miRBase ID: ptr-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-221
          Stem-loop Accession # MI0040329
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049559
          miRBase ID: pha-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-221
          Stem-loop Accession # MI0002894
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002585
          miRBase ID: ppy-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-221-1
          Stem-loop Accession # MI0034851
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUGGUUCAAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042889
          miRBase ID: pny-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Stem-loop ID pny-mir-221-2
          Stem-loop Accession # MI0034858
          Stem-loop Sequence
          ACCUGGCAUACAGUGUAGGAAUCUGUGUGUGUCAGUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042889
          miRBase ID: pny-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-221
          Stem-loop Accession # MI0039357
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUUUUGGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048355
          miRBase ID: ocu-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-221
          Stem-loop Accession # MI0002892
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002583
          miRBase ID: mml-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-221
          Stem-loop Accession # MI0040239
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049481
          miRBase ID: sbo-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-221
          Stem-loop Accession # MI0013710
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAAGCAACAGCUACAUUGUCUGCUGGGUUUCCAGCUGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014512
          miRBase ID: tgu-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-221
          Stem-loop Accession # MI0003222
          Stem-loop Sequence
          CCUGAACCUGGCAUACAAUGUAGAUUUCUGUGUGCUUGGAUUCUACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002907
          miRBase ID: tni-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-221
          Stem-loop Accession # MI0004953
          Stem-loop Sequence
          ACCCAGACUUUGGCGUGCACCUGGCAUACAAUGUAGAAAACUGUGUUUGCAAAGCAACAGCUACAUUGUCUGCUGGGUUUCAUGCUGAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003700
          miRBase ID: xtr-miR-221
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-221
          Stem-loop Accession # MI0001387
          Stem-loop Sequence
          GUCGUGAACCUGGCAUACAAUGUAGAUUUCUGUGUGGUACUAUCUACAGCUACAUUGUCUGCUGGGUUUCAGGCCAGCAGAAUAAUUCUGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001288
          miRBase ID: dre-miR-221-3p
          Mature miRNA Sequence: AGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465464_pri
          TaqMan™ Pri-miRNA Assay : Mm03307181_pri
          TaqMan™ Pri-miRNA Assay : Hs03303007_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10337
          Ambion® Pre-miR™ miRNA Precursor : PM10337
          TaqMan™ MicroRNA Assay : 000524
          mirVana® miRNA inhibitor : MH10337
          mirVana® miRNA mimic : MC10337
          TaqMan™ Advanced miRNA Assay : mmu481005_mir
          TaqMan™ Advanced miRNA Assay : 477981_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline