Search
Search

| Assay Name: | hsa-miR-221-3p |
|---|---|
| miRBase Accession Number: | MI0000298 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCUACAUUGUCUGCUGGGUUUC |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Chicken, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Fugu rubripes, Gorilla gorilla, Guinea pig, Horse, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Oryzias latipes, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pundamilia nyererei, Rabbit, Rhesus monkey, Saimiri boliviensis, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | AGCUACAUUGUCUGCUGGGUUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-221-3p | MIMAT0000278 |
| Mouse | mmu-miR-221-3p | MIMAT0000669 |
| Rat | rno-miR-221-3p | MIMAT0000890 |
| Alligator mississippiensis | ami-miR-221-3p | MIMAT0038253 |
| Astatotilapia burtoni | abu-miR-221 | MIMAT0042073 |
| Chicken | gga-miR-221-3p | MIMAT0001108 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-221-3p | MIMAT0023859 |
| Dasypus novemcinctus | dno-miR-221-3p | MIMAT0047776 |
| Daubentonia madagascariensis | dma-miR-221 | MIMAT0049278 |
| Fugu rubripes | fru-miR-221 | MIMAT0002906 |
| Gorilla gorilla | ggo-miR-221 | MIMAT0002584 |
| Guinea pig | cpo-miR-221-3p | MIMAT0047131 |
| Horse | eca-miR-221 | MIMAT0013203 |
| Metriaclima zebra | mze-miR-221 | MIMAT0042316 |
| Microcebus murinus | mmr-miR-221 | MIMAT0049144 |
| Monodelphis domestica | mdo-miR-221-3p | MIMAT0004160 |
| Neolamprologus brichardi | nbr-miR-221 | MIMAT0042495 |
| Nomascus leucogenys | nle-miR-221 | MIMAT0049404 |
| Ophiophagus hannah | oha-miR-221-3p | MIMAT0036847 |
| Oreochromis niloticus | oni-miR-221 | MIMAT0042675 |
| Oryzias latipes | ola-miR-221 | MIMAT0022530 |
| Otolemur garnettii | oga-miR-221 | MIMAT0049658 |
| Pan paniscus | ppa-miR-221 | MIMAT0002586 |
| Pan troglodytes | ptr-miR-221 | MIMAT0008076 |
| Papio hamadryas | pha-miR-221 | MIMAT0049559 |
| Pongo pygmaeus | ppy-miR-221 | MIMAT0002585 |
| Pundamilia nyererei | pny-miR-221 | MIMAT0042889 |
| Rabbit | ocu-miR-221-3p | MIMAT0048355 |
| Rhesus monkey | mml-miR-221-3p | MIMAT0002583 |
| Saimiri boliviensis | sbo-miR-221 | MIMAT0049481 |
| Taeniopygia guttata | tgu-miR-221-3p | MIMAT0014512 |
| Tetraodon nigroviridis | tni-miR-221 | MIMAT0002907 |
| Xenopus tropicalis | xtr-miR-221 | MIMAT0003700 |
| Zebrafish | dre-miR-221-3p | MIMAT0001288 |