Search
Search

| Assay Name: | rno-miR-374-5p |
|---|---|
| Stem-loop Accession Number: | MI0005566 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUAUAAUACAACCUGCUAAGUG |
| Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Dog, Goat, Gorilla gorilla, Horse, Pan troglodytes, Pig, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | AUAUAAUACAACCUGCUAAGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374b-5p | MIMAT0004955 |
| Mouse | mmu-miR-374b-5p | MIMAT0003727 |
| Rat | rno-miR-374-5p | MIMAT0003208 |
| Bovine | bta-miR-374b | MIMAT0009302 |
| Cricetulus griseus | cgr-miR-374-5p | MIMAT0023939 |
| Dog | cfa-miR-374b | MIMAT0006754 |
| View More Rows | ||
| Goat | chi-miR-374b-5p | MIMAT0036182 |
| Gorilla gorilla | ggo-miR-374b | MIMAT0024126 |
| Horse | eca-miR-374b | MIMAT0013214 |
| Pan troglodytes | ptr-miR-374b | MIMAT0008124 |
| Pig | ssc-miR-374b-5p | MIMAT0013915 |
| Pteropus alecto | pal-miR-374b-5p | MIMAT0039989 |
| Rabbit | ocu-miR-374b-5p | MIMAT0048408 |
| Rhesus monkey | mml-miR-374b-5p | MIMAT0006301 |