Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11339
          Assay Name: hsa-miR-374b-5p
          miRBase Accession Number: MI0005566
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 74218547 - 74218618 [-] on Build GRCh38
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Dog, Goat, Gorilla gorilla, Horse, Pan troglodytes, Pig, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC11339

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000288, mir-374

          Mature miRNA Sequence:

          AUAUAAUACAACCUGCUAAGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-374b-5p MIMAT0004955
          Mouse mmu-miR-374b-5p MIMAT0003727
          Rat rno-miR-374-5p MIMAT0003208
          Bovine bta-miR-374b MIMAT0009302
          Cricetulus griseus cgr-miR-374-5p MIMAT0023939
          Dog cfa-miR-374b MIMAT0006754
          View More Rows
          Goat chi-miR-374b-5p MIMAT0036182
          Gorilla gorilla ggo-miR-374b MIMAT0024126
          Horse eca-miR-374b MIMAT0013214
          Pan troglodytes ptr-miR-374b MIMAT0008124
          Pig ssc-miR-374b-5p MIMAT0013915
          Pteropus alecto pal-miR-374b-5p MIMAT0039989
          Rabbit ocu-miR-374b-5p MIMAT0048408
          Rhesus monkey mml-miR-374b-5p MIMAT0006301

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-374b
          Stem-loop Accession # MI0005566
          Stem-loop Sequence
          ACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAGCAGGUUGUAUUAUCAUUGUCCGUGUCU
          Chromosome Location Chr. X - 74218547 - 74218618 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004955
          miRBase ID: hsa-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: Chr. X - 74218547 - 74218618 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-374b
          Stem-loop Accession # MI0004125
          Stem-loop Sequence
          GAAGAAAUCCUACUCGGGUGGAUAUAAUACAACCUGCUAAGUGUUCUAGCACUUAGCAGGUUGUAUUAUCAUUGUCCGAGGUUAUGGCUCUCGUC
          Chromosome Location Chr. X - 103573060 - 103573154 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003727
          miRBase ID: mmu-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: Chr. X - 103573060 - 103573154 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-374
          Stem-loop Accession # MI0003552
          Stem-loop Sequence
          CUCGGAUGGAUAUAAUACAACCUGCUAAGUGUUCUAGCACUUAGCACGUUGUAUUAUUAUUGUCCGAG
          Chromosome Location Chr. X - 74443648 - 74443715 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003208
          miRBase ID: rno-miR-374-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: Chr. X - 74443648 - 74443715 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-374b
          Stem-loop Accession # MI0009816
          Stem-loop Sequence
          GAAGAAAUCCUACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAUCAGGUUGUAUUAUCAUUGUCCGUGUCUAUGGCUCUCGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009302
          miRBase ID: bta-miR-374b
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-374
          Stem-loop Accession # MI0020513
          Stem-loop Sequence
          CUCGGAUGGAUAUAAUACAACCUGCUAAGUGUUCUAGCACUUAUCAGGUUGUAUUAUCAUUGUCCGUGUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023939
          miRBase ID: cgr-miR-374-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-374b
          Stem-loop Accession # MI0008180
          Stem-loop Sequence
          AUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAUCAGGUUGUAUUAUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006754
          miRBase ID: cfa-miR-374b
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-374b
          Stem-loop Accession # MI0030766
          Stem-loop Sequence
          GAAGAAAUCCUACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAUCAGGUUGUAUUAUCAUUGUCCGUGUCUAUGGCUCUCGUCUACCAGACUCUAAAUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036182
          miRBase ID: chi-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-374b
          Stem-loop Accession # MI0020673
          Stem-loop Sequence
          AAGAAAUCCUACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAGCAGGUUGUAUUAUCAUUGUCCGUGUCUAUGGCUCUCGUCUACCAGACUUUAAAUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024126
          miRBase ID: ggo-miR-374b
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-374b
          Stem-loop Accession # MI0012960
          Stem-loop Sequence
          ACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAUCAGGUUGUAUUAUCAUUGUCCGUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013214
          miRBase ID: eca-miR-374b
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-374b
          Stem-loop Accession # MI0008644
          Stem-loop Sequence
          ACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAGCAGGUUGUAUUAUCAUUGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008124
          miRBase ID: ptr-miR-374b
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-374b
          Stem-loop Accession # MI0013131
          Stem-loop Sequence
          UCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAUCAGGUUGUAUUAUCAUUGUCCGUGUCUAUGGCUCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013915
          miRBase ID: ssc-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-374b
          Stem-loop Accession # MI0032436
          Stem-loop Sequence
          AUAUAAUACAACCUGCUAAGUGUCCCAGCACUUAGCAGGUUGUAUUAUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039989
          miRBase ID: pal-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-374b
          Stem-loop Accession # MI0039385
          Stem-loop Sequence
          AUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAGCAGGUUGUAUUAUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048408
          miRBase ID: ocu-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-374b
          Stem-loop Accession # MI0007719
          Stem-loop Sequence
          ACUCGGAUGGAUAUAAUACAACCUGCUAAGUGUCCUAGCACUUAGCAGGUUGUAUUAUCAUUGUCCGUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006301
          miRBase ID: mml-miR-374b-5p
          Mature miRNA Sequence: AUAUAAUACAACCUGCUAAGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465535_pri
          TaqMan™ Pri-miRNA Assay : Hs03295671_pri
          TaqMan™ Pri-miRNA Assay : Mm03308494_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11339
          Ambion® Pre-miR™ miRNA Precursor : PM11339
          TaqMan™ MicroRNA Assay : 001319
          mirVana® miRNA inhibitor : MH11339
          TaqMan™ Advanced miRNA Assay : rno481140_mir
          TaqMan™ Advanced miRNA Assay : mmu481140_mir
          TaqMan™ Advanced miRNA Assay : 478389_mir


          miRBase Alias:

          Human: hsa-miR-374b (v17)
          Mouse: mmu-miR-374-5p (v18)
          Mouse: mmu-miR-374 (v17)
          Rat: rno-miR-374 (v18)
          Macaca mulatta: mml-miR-374b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline