Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481141_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-375-3p
          Stem-loop Accession Number: MI0000783
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 219001645 - 219001708 [-] on Build GRCh38
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Species: Human, Mouse, Rat, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481141_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000114, mir-375

          Mature miRNA Sequence:

          UUUGUUCGUUCGGCUCGCGUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-375-3p MIMAT0000728
          Mouse mmu-miR-375-3p MIMAT0000739
          Rat rno-miR-375-3p MIMAT0005307
          Dasypus novemcinctus dno-miR-375-3p MIMAT0047849
          Dog cfa-miR-375 MIMAT0009871
          Gorilla gorilla ggo-miR-375 MIMAT0024189
          View More Rows
          Guinea pig cpo-miR-375-3p MIMAT0047208
          Pan troglodytes ptr-miR-375 MIMAT0008125
          Pig ssc-miR-375 MIMAT0041608
          Pongo pygmaeus ppy-miR-375 MIMAT0015863
          Pteropus alecto pal-miR-375-3p MIMAT0040121
          Rabbit ocu-miR-375-3p MIMAT0048411
          Rhesus monkey mml-miR-375 MIMAT0006302

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-375
          Stem-loop Accession # MI0000783
          Stem-loop Sequence
          CCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGC
          Chromosome Location Chr. 2 - 219001645 - 219001708 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000728
          miRBase ID: hsa-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: Chr. 2 - 219001645 - 219001708 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-375
          Stem-loop Accession # MI0000792
          Stem-loop Sequence
          CCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGC
          Chromosome Location Chr. 1 - 74900658 - 74900721 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000739
          miRBase ID: mmu-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: Chr. 1 - 74900658 - 74900721 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-375
          Stem-loop Accession # MI0006140
          Stem-loop Sequence
          UCCGGCCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGCAGGGGC
          Chromosome Location Chr. 9 - 82161716 - 82161790 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005307
          miRBase ID: rno-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: Chr. 9 - 82161716 - 82161790 [-] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-375
          Stem-loop Accession # MI0039089
          Stem-loop Sequence
          GCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047849
          miRBase ID: dno-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-375
          Stem-loop Accession # MI0010368
          Stem-loop Sequence
          GCCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGCAGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009871
          miRBase ID: cfa-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-375
          Stem-loop Accession # MI0020736
          Stem-loop Sequence
          CGACCGCCAGGCCCCGGGCCCUCCGCUCCCGCCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGCAGGGGCGGCCUCUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024189
          miRBase ID: ggo-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-375
          Stem-loop Accession # MI0038752
          Stem-loop Sequence
          GCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047208
          miRBase ID: cpo-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-375
          Stem-loop Accession # MI0008645
          Stem-loop Sequence
          CCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008125
          miRBase ID: ptr-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-375
          Stem-loop Accession # MI0033409
          Stem-loop Sequence
          CCCCCGCCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGCAGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041608
          miRBase ID: ssc-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-375
          Stem-loop Accession # MI0014920
          Stem-loop Sequence
          CCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015863
          miRBase ID: ppy-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-375
          Stem-loop Accession # MI0032504
          Stem-loop Sequence
          GCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040121
          miRBase ID: pal-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-375
          Stem-loop Accession # MI0039386
          Stem-loop Sequence
          GCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048411
          miRBase ID: ocu-miR-375-3p
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-375
          Stem-loop Accession # MI0007720
          Stem-loop Sequence
          CCCCGCGACGAGCCCCUCGCACAAACCGGACCUGAGCGUUUUGUUCGUUCGGCUCGCGUGAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006302
          miRBase ID: mml-miR-375
          Mature miRNA Sequence: UUUGUUCGUUCGGCUCGCGUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307340_pri
          TaqMan™ Pri-miRNA Assay : Hs03303515_pri
          TaqMan™ Pri-miRNA Assay : Rn03465608_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10327
          Ambion® Pre-miR™ miRNA Precursor : PM10327
          TaqMan™ MicroRNA Assay : 000564
          mirVana® miRNA inhibitor : MH10327
          mirVana® miRNA mimic : MC10327
          TaqMan™ Advanced miRNA Assay : mmu481141_mir
          TaqMan™ Advanced miRNA Assay : 478074_mir


          miRBase Alias:

          Human: hsa-miR-375 (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline