Search
Search

| Assay Name: | hsa-miR-375-3p |
|---|---|
| miRBase Accession Number: | MI0000783 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGUUCGUUCGGCUCGCGUGA |
| Species: | Human, Mouse, Rat, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UUUGUUCGUUCGGCUCGCGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-375-3p | MIMAT0000728 |
| Mouse | mmu-miR-375-3p | MIMAT0000739 |
| Rat | rno-miR-375-3p | MIMAT0005307 |
| Dasypus novemcinctus | dno-miR-375-3p | MIMAT0047849 |
| Dog | cfa-miR-375 | MIMAT0009871 |
| Gorilla gorilla | ggo-miR-375 | MIMAT0024189 |
| View More Rows | ||
| Guinea pig | cpo-miR-375-3p | MIMAT0047208 |
| Pan troglodytes | ptr-miR-375 | MIMAT0008125 |
| Pig | ssc-miR-375 | MIMAT0041608 |
| Pongo pygmaeus | ppy-miR-375 | MIMAT0015863 |
| Pteropus alecto | pal-miR-375-3p | MIMAT0040121 |
| Rabbit | ocu-miR-375-3p | MIMAT0048411 |
| Rhesus monkey | mml-miR-375 | MIMAT0006302 |