Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481149_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-382-5p
          Stem-loop Accession Number: MI0000790
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101054306 - 101054381 [+] on Build GRCh38
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Horse, Pan troglodytes, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481149_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          GAAGUUGUUCGUGGUGGAUUCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-382-5p MIMAT0000737
          Mouse mmu-miR-382-5p MIMAT0000747
          Rat rno-miR-382-5p MIMAT0003201
          Bovine bta-miR-382 MIMAT0009308
          Callithrix jacchus cja-miR-382 MIMAT0039409
          Cricetulus griseus cgr-miR-382 MIMAT0023947
          View More Rows
          Dasypus novemcinctus dno-miR-382-5p MIMAT0047866
          Goat chi-miR-382-5p MIMAT0036200
          Guinea pig cpo-miR-382-5p MIMAT0047227
          Horse eca-miR-382 MIMAT0013150
          Pan troglodytes ptr-miR-382 MIMAT0008134
          Pteropus alecto pal-miR-382-5p MIMAT0039915
          Rabbit ocu-miR-382-5p MIMAT0048427
          Rhesus monkey mml-miR-382-5p MIMAT0006311
          Sheep oar-miR-382-5p MIMAT0019306

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-382
          Stem-loop Accession # MI0000790
          Stem-loop Sequence
          UACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location Chr. 14 - 101054306 - 101054381 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000737
          miRBase ID: hsa-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: Chr. 14 - 101054306 - 101054381 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-382
          Stem-loop Accession # MI0000799
          Stem-loop Sequence
          UACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUGUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location Chr. 12 - 109733771 - 109733846 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000747
          miRBase ID: mmu-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: Chr. 12 - 109733771 - 109733846 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-382
          Stem-loop Accession # MI0003548
          Stem-loop Sequence
          GGUACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUGUGACGAAUCAUUCACGGACAACACUUUUUUCAGUACC
          Chromosome Location Chr. 6 - 133884178 - 133884257 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003201
          miRBase ID: rno-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: Chr. 6 - 133884178 - 133884257 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-382
          Stem-loop Accession # MI0009822
          Stem-loop Sequence
          UACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009308
          miRBase ID: bta-miR-382
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-382
          Stem-loop Accession # MI0031929
          Stem-loop Sequence
          GCCGUGGUACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUACCAAAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039409
          miRBase ID: cja-miR-382
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-382
          Stem-loop Accession # MI0020518
          Stem-loop Sequence
          ACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUGUGACGAAUCAUUCACGGACAACACUUUUUUCAGUACCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023947
          miRBase ID: cgr-miR-382
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-382
          Stem-loop Accession # MI0039098
          Stem-loop Sequence
          GAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047866
          miRBase ID: dno-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-382
          Stem-loop Accession # MI0030777
          Stem-loop Sequence
          CCCUGCUCUGUCUUGCCUCUGCCUUUCUGUGGUACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUACCAAAUACUACCUCUAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036200
          miRBase ID: chi-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-382
          Stem-loop Accession # MI0038762
          Stem-loop Sequence
          GAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047227
          miRBase ID: cpo-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-382
          Stem-loop Accession # MI0012895
          Stem-loop Sequence
          UACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013150
          miRBase ID: eca-miR-382
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-382
          Stem-loop Accession # MI0008655
          Stem-loop Sequence
          ACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008134
          miRBase ID: ptr-miR-382
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-382
          Stem-loop Accession # MI0032398
          Stem-loop Sequence
          GAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039915
          miRBase ID: pal-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-382
          Stem-loop Accession # MI0039395
          Stem-loop Sequence
          GAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048427
          miRBase ID: ocu-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-382
          Stem-loop Accession # MI0007730
          Stem-loop Sequence
          UACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006311
          miRBase ID: mml-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-382
          Stem-loop Accession # MI0016947
          Stem-loop Sequence
          CCCUGCUCUGUCUUGCCUCUGCCUUUCUGUGGUACUUGAAGAGAAGUUGUUCGUGGUGGAUUCGCUUUACUUAUGACGAAUCAUUCACGGACAACACUUUUUUCAGUACCAAAUACUACCUCUAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019306
          miRBase ID: oar-miR-382-5p
          Mature miRNA Sequence: GAAGUUGUUCGUGGUGGAUUCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307385_pri
          TaqMan™ Pri-miRNA Assay : Hs03303562_pri
          TaqMan™ Pri-miRNA Assay : Rn03465298_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10924
          Ambion® Pre-miR™ miRNA Precursor : PM10924
          TaqMan™ MicroRNA Assay : 000572
          mirVana® miRNA inhibitor : MH10924
          mirVana® miRNA mimic : MC10924
          TaqMan™ Advanced miRNA Assay : 478078_mir
          TaqMan™ Advanced miRNA Assay : mmu481149_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline