Search
Search

| Assay Name: | hsa-miR-382-5p |
|---|---|
| miRBase Accession Number: | MI0000790 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAAGUUGUUCGUGGUGGAUUCG |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Horse, Pan troglodytes, Pteropus alecto, Rabbit, Rhesus monkey, Sheep |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GAAGUUGUUCGUGGUGGAUUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-382-5p | MIMAT0000737 |
| Mouse | mmu-miR-382-5p | MIMAT0000747 |
| Rat | rno-miR-382-5p | MIMAT0003201 |
| Bovine | bta-miR-382 | MIMAT0009308 |
| Callithrix jacchus | cja-miR-382 | MIMAT0039409 |
| Cricetulus griseus | cgr-miR-382 | MIMAT0023947 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-382-5p | MIMAT0047866 |
| Goat | chi-miR-382-5p | MIMAT0036200 |
| Guinea pig | cpo-miR-382-5p | MIMAT0047227 |
| Horse | eca-miR-382 | MIMAT0013150 |
| Pan troglodytes | ptr-miR-382 | MIMAT0008134 |
| Pteropus alecto | pal-miR-382-5p | MIMAT0039915 |
| Rabbit | ocu-miR-382-5p | MIMAT0048427 |
| Rhesus monkey | mml-miR-382-5p | MIMAT0006311 |
| Sheep | oar-miR-382-5p | MIMAT0019306 |