Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481350_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-23a-5p
          Stem-loop Accession Number: MI0000079
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 13836587 - 13836659 [-] on Build GRCh38
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Species: Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481350_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000027, mir-23

          Mature miRNA Sequence:

          GGGGUUCCUGGGGAUGGGAUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-23a-5p MIMAT0004496
          Mouse mmu-miR-23a-5p MIMAT0017019
          Rat rno-miR-23a-5p MIMAT0004712
          Cricetulus griseus cgr-miR-23a-5p MIMAT0023862
          Dasypus novemcinctus dno-miR-23a-5p MIMAT0047528
          Guinea pig cpo-miR-23a-5p MIMAT0046876

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-23a
          Stem-loop Accession # MI0000079
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004496
          miRBase ID: hsa-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-23a
          Stem-loop Accession # MI0000571
          Stem-loop Sequence
          CGGACGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCC
          Chromosome Location Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017019
          miRBase ID: mmu-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-23a
          Stem-loop Accession # MI0000852
          Stem-loop Sequence
          CGGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCC
          Chromosome Location Chr. 19 - 25318582 - 25318656 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004712
          miRBase ID: rno-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: Chr. 19 - 25318582 - 25318656 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-23a
          Stem-loop Accession # MI0020466
          Stem-loop Sequence
          UUGGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023862
          miRBase ID: cgr-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-23a
          Stem-loop Accession # MI0038919
          Stem-loop Sequence
          GGGGUUCCUGGGGAUGGGAUUUGCUGUCGGUCACAAAUCACAUUGCCAGGGAUUUCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047528
          miRBase ID: dno-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-23a
          Stem-loop Accession # MI0038575
          Stem-loop Sequence
          GGGGUUCCUGGGGAUGGGAUUUGCUGUCUGUCACAAAUCACAUUGCCAGGGAUUUCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046876
          miRBase ID: cpo-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04270764_pri
          TaqMan™ Pri-miRNA Assay : Rn03465976_pri
          TaqMan™ Pri-miRNA Assay : Mm03306867_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12940
          Ambion® Pre-miR™ miRNA Precursor : PM12940
          TaqMan™ MicroRNA Assay : 002439
          mirVana® miRNA inhibitor : MH12940
          mirVana® miRNA mimic : MC12940
          TaqMan™ Advanced miRNA Assay : 478782_mir
          TaqMan™ Advanced miRNA Assay : mmu482713_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline