Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04270764_pri
          Assay Name:
          hsa-mir-23a
          Stem-loop ID:
          hsa-mir-23a hsa-mir-24-2 hsa-mir-27a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.19: 13836587 - 13836659 [-] on Build GRCh38
          Stem-loop Location:
          Chr.19: 13836587 - 13836659 [-] on Build GRCh38 Chr.19: 13836287 - 13836359 [-] on Build GRCh38 Chr.19: 13836440 - 13836517 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-23a-5p hsa-miR-23a-3p hsa-miR-24-3p hsa-miR-24-2-5p hsa-miR-27a-5p hsa-miR-27a-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs04270764_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          65

          Chromosome Location:

          Chr. 19 - 13836805 - 13836805 [+] on 13836805 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-23a-5p MIMAT0004496
          Human hsa-miR-23a-3p MIMAT0000078
          Human hsa-miR-24-3p MIMAT0000080
          Human hsa-miR-24-2-5p MIMAT0004497
          Human hsa-miR-27a-5p MIMAT0004501
          Human hsa-miR-27a-3p MIMAT0000084

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-23a
          Stem-loop Accession # MI0000079
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004496
          miRBase ID: hsa-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38

          miRBase Accession #: MIMAT0000078
          miRBase ID: hsa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38

          Gene Family ID MIPF0000027, mir-23
          Distance to Amplicon 108 bases
          Stem-loop ID hsa-mir-24-2
          Stem-loop Accession # MI0000081
          Stem-loop Sequence
          CUCUGCCUCCCGUGCCUACUGAGCUGAAACACAGUUGGUUUGUGUACACUGGCUCAGUUCAGCAGGAACAGGG
          Chromosome Location Chr. 19 - 13836287 - 13836359 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000080
          miRBase ID: hsa-miR-24-3p
          Mature miRNA Sequence: UGGCUCAGUUCAGCAGGAACAG
          Chromosome Location: Chr. 19 - 13836287 - 13836359 [-] on Build GRCh38

          miRBase Accession #: MIMAT0004497
          miRBase ID: hsa-miR-24-2-5p
          Mature miRNA Sequence: UGCCUACUGAGCUGAAACACAG
          Chromosome Location: Chr. 19 - 13836287 - 13836359 [-] on Build GRCh38

          Gene Family ID MIPF0000041, mir-24
          Distance to Amplicon 408 bases
          Stem-loop ID hsa-mir-27a
          Stem-loop Accession # MI0000085
          Stem-loop Sequence
          CUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUCCACACCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCAG
          Chromosome Location Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004501
          miRBase ID: hsa-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38

          miRBase Accession #: MIMAT0000084
          miRBase ID: hsa-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38

          Gene Family ID MIPF0000036, mir-27
          Distance to Amplicon 250 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM13096
          Ambion® Pre-miR™ miRNA Precursor : PM13096
          TaqMan™ MicroRNA Assay : 002445
          mirVana® miRNA inhibitor : MH13096
          mirVana® miRNA mimic : MC13096
          TaqMan™ Advanced miRNA Assay : rno481015_mir
          TaqMan™ Advanced miRNA Assay : 477998_mir
          TaqMan™ Advanced miRNA Assay : mmu481015_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10939
          Ambion® Pre-miR™ miRNA Precursor : PM10939
          TaqMan™ MicroRNA Assay : 000408
          mirVana® miRNA inhibitor : MH10939
          mirVana® miRNA mimic : MC10939
          TaqMan™ Advanced miRNA Assay : 478384_mir
          TaqMan™ Advanced miRNA Assay : mmu478384_mir
          TaqMan™ Advanced miRNA Assay : rno478384_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12940
          Ambion® Pre-miR™ miRNA Precursor : PM12940
          TaqMan™ MicroRNA Assay : 002439
          mirVana® miRNA inhibitor : MH12940
          mirVana® miRNA mimic : MC12940
          TaqMan™ Advanced miRNA Assay : 478782_mir
          TaqMan™ Advanced miRNA Assay : mmu482713_mir
          TaqMan™ Advanced miRNA Assay : rno481350_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10644
          Ambion® Pre-miR™ miRNA Precursor : PM10644
          TaqMan™ MicroRNA Assay : 000399
          mirVana® miRNA inhibitor : MH10644
          mirVana® miRNA mimic : MC10644
          TaqMan™ Advanced miRNA Assay : 478532_mir
          TaqMan™ Advanced miRNA Assay : mmu478532_mir
          TaqMan™ Advanced miRNA Assay : rno478532_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10737
          Ambion® Pre-miR™ miRNA Precursor : PM10737
          TaqMan™ MicroRNA Assay : 000402
          mirVana® miRNA inhibitor : MH10737
          mirVana® miRNA mimic : MC10737
          TaqMan™ Advanced miRNA Assay : 477992_mir
          TaqMan™ Advanced miRNA Assay : rno481011_mir
          TaqMan™ Advanced miRNA Assay : mmu481011_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12498
          Ambion® Pre-miR™ miRNA Precursor : PM12498
          TaqMan™ MicroRNA Assay : 002441
          mirVana® miRNA inhibitor : MH12498
          mirVana® miRNA mimic : MC12498
          TaqMan™ Advanced miRNA Assay : 478785_mir


          miRBase Alias:

          Human: hsa-mir-24-2 (v21)
          Human: hsa-mir-27a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline