Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10644
          Assay Name: hsa-miR-23a-3p
          miRBase Accession Number: MI0000079
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 13836587 - 13836659 [-] on Build GRCh38
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Species: Human, Mouse, Rat, Anolis carolinensis, Ateles geoffroyi, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Goat, Gorilla gorilla, Horse, Ictalurus punctatus, Lemur catta, Macaca nemestrina, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pundamilia nyererei, Python bivittatus, Rhesus monkey, Saguinus labiatus, Salmo salar, Xenopus laevis, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10644

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000027, mir-23

          Mature miRNA Sequence:

          AUCACAUUGCCAGGGAUUUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-23a-3p MIMAT0000078
          Mouse mmu-miR-23a-3p MIMAT0000532
          Rat rno-miR-23a-3p MIMAT0000792
          Anolis carolinensis aca-miR-23a-3p MIMAT0021896
          Ateles geoffroyi age-miR-23a MIMAT0002741
          Chrysemys picta cpi-miR-23a-3p MIMAT0037655
          View More Rows
          Cricetulus griseus cgr-miR-23a-3p MIMAT0023863
          Cyprinus carpio ccr-miR-23a MIMAT0026277
          Goat chi-miR-23a MIMAT0036087
          Gorilla gorilla ggo-miR-23a MIMAT0002738
          Horse eca-miR-23a MIMAT0012986
          Ictalurus punctatus ipu-miR-23a MIMAT0029481
          Lemur catta lca-miR-23a MIMAT0002745
          Macaca nemestrina mne-miR-23a MIMAT0002756
          Pan paniscus ppa-miR-23a MIMAT0002743
          Pan troglodytes ptr-miR-23a MIMAT0002749
          Papio hamadryas pha-miR-23a MIMAT0049533
          Pig ssc-miR-23a MIMAT0002133
          Pongo pygmaeus ppy-miR-23a MIMAT0002747
          Pundamilia nyererei pny-miR-23a MIMAT0042955
          Python bivittatus pbv-miR-23a-3p MIMAT0038853
          Rhesus monkey mml-miR-23a-3p MIMAT0002752
          Saguinus labiatus sla-miR-23a MIMAT0002754
          Salmo salar ssa-miR-23a-3p MIMAT0032549
          Xenopus laevis xla-miR-23a MIMAT0011145
          Xenopus tropicalis xtr-miR-23a MIMAT0003568

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-23a
          Stem-loop Accession # MI0000079
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000078
          miRBase ID: hsa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-23a
          Stem-loop Accession # MI0000571
          Stem-loop Sequence
          CGGACGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCC
          Chromosome Location Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000532
          miRBase ID: mmu-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-23a
          Stem-loop Accession # MI0000852
          Stem-loop Sequence
          CGGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCC
          Chromosome Location Chr. 19 - 25318582 - 25318656 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000792
          miRBase ID: rno-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: Chr. 19 - 25318582 - 25318656 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-23a
          Stem-loop Accession # MI0018831
          Stem-loop Sequence
          CCGCUGCCAGCUGGGGUUCCUGGUGAUGUGAUUUUAUUUUGGUGUUCAGAAUCACAUUGCCAGGGAUUUCCAACCAUCAGUGAGCUACCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021896
          miRBase ID: aca-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-23a
          Stem-loop Accession # MI0003038
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUGCCUAUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002741
          miRBase ID: age-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-23a
          Stem-loop Accession # MI0029411
          Stem-loop Sequence
          CCUGCCCCUGCCAGCUGGGGUUCCUGGUGAUGCGAUUUUUAUCCUACCACCCAAAAUCACAUUGCCAGGGAUUUCCAACGGGCGGCGGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037655
          miRBase ID: cpi-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-23a
          Stem-loop Accession # MI0020466
          Stem-loop Sequence
          UUGGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023863
          miRBase ID: cgr-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-23a
          Stem-loop Accession # MI0023378
          Stem-loop Sequence
          CCAGCUGGAGGGAUUCCUGGCAGAGUGAUUUGGGAUUAUAUCAUAAAAUCACAUUGCCAGGGAUUUCCAACCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026277
          miRBase ID: ccr-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-23a
          Stem-loop Accession # MI0030711
          Stem-loop Sequence
          UGGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUGCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036087
          miRBase ID: chi-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-23a
          Stem-loop Accession # MI0003035
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002738
          miRBase ID: ggo-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-23a
          Stem-loop Accession # MI0012737
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUGCUUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012986
          miRBase ID: eca-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-23a-1
          Stem-loop Accession # MI0024593
          Stem-loop Sequence
          GGGUUCCUGGCACCGUGAUUUGGUGGAUAAACAGAAAUGAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029481
          miRBase ID: ipu-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-23a-2
          Stem-loop Accession # MI0024591
          Stem-loop Sequence
          GGAAUUCCUGGCAGAGUGAUUUUUAAACCUAAUGACUGAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029481
          miRBase ID: ipu-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-23a-3
          Stem-loop Accession # MI0024592
          Stem-loop Sequence
          GGAUUCCUGGCAGAGUGAUUUGGGAUUAUAUCAUAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029481
          miRBase ID: ipu-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Lemur catta

          Stem-loop ID lca-mir-23a
          Stem-loop Accession # MI0003043
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUACCUAUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002745
          miRBase ID: lca-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-23a
          Stem-loop Accession # MI0003056
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002756
          miRBase ID: mne-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-23a
          Stem-loop Accession # MI0003040
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002743
          miRBase ID: ppa-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-23a
          Stem-loop Accession # MI0003048
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002749
          miRBase ID: ptr-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-23a
          Stem-loop Accession # MI0040299
          Stem-loop Sequence
          GGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049533
          miRBase ID: pha-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-23a
          Stem-loop Accession # MI0002427
          Stem-loop Sequence
          CGGCUGGGGUUCCUGGGGAUGGGAUUUGCUGCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAAUCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002133
          miRBase ID: ssc-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-23a
          Stem-loop Accession # MI0003045
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002747
          miRBase ID: ppy-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-23a-1
          Stem-loop Accession # MI0034942
          Stem-loop Sequence
          GAAUUCCUGGCAGAGUGAUUUUUUUUUAACUAGAGGACUGAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042955
          miRBase ID: pny-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID pny-mir-23a-2
          Stem-loop Accession # MI0034946
          Stem-loop Sequence
          GGGUUCCUGGCACCGUGAUUUGAUGCACAAAGAGAAACAAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042955
          miRBase ID: pny-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID pny-mir-23a-3
          Stem-loop Accession # MI0034949
          Stem-loop Sequence
          GGAUUCCUGGCAGAGUGAUUUGGUUGUGAUAUAAUGUAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042955
          miRBase ID: pny-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-23a
          Stem-loop Accession # MI0030170
          Stem-loop Sequence
          GCUGCCUGCUGGGGUUCCUGGUGAUGUGAUUUUACCCUGAUGUUCAGAAUCACAUUGCCAGGGAUUUCCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038853
          miRBase ID: pbv-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-23a
          Stem-loop Accession # MI0003051
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002752
          miRBase ID: mml-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-23a
          Stem-loop Accession # MI0003054
          Stem-loop Sequence
          GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUGCCUAUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002754
          miRBase ID: sla-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-23a-1
          Stem-loop Accession # MI0026638
          Stem-loop Sequence
          GGAUUCCUGGCAGAGUGAUUUGAUAUUGUAUCAUAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032549
          miRBase ID: ssa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-23a-2
          Stem-loop Accession # MI0026639
          Stem-loop Sequence
          GGAUUCCUGGCAGAGUGAUUUGGUAUUGUGUCAUAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032549
          miRBase ID: ssa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-23a-3
          Stem-loop Accession # MI0026636
          Stem-loop Sequence
          GGAAUUCCUGGUAGAGUGAUUUUUAAGACUAAAGACUGAAUCACAUUGCCAGGGAUUUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032549
          miRBase ID: ssa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-23a-4
          Stem-loop Accession # MI0026637
          Stem-loop Sequence
          AGGGUUCCUGGCACAGUGAUUUGGUGCACAAAUAUAAACGAAAAUCACAUUGCCAGGGAUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032549
          miRBase ID: ssa-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-23a
          Stem-loop Accession # MI0008383
          Stem-loop Sequence
          CCGACAGCGGGGGAUUCCUGGAGAUGGGAUUUUAUUUUUGCAGCCAAUAAAUCACAUUGCCAGGGAUUUCCAACUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011145
          miRBase ID: xla-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-23a-1
          Stem-loop Accession # MI0004894
          Stem-loop Sequence
          CUGACAGCGGGGGAUUCCUGGAGAUGGGAUUUUAUUUUGGCAGCCAAUAAAUCACAUUGCCAGGGAUUUCCAACUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003568
          miRBase ID: xtr-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA

          Stem-loop ID xtr-mir-23a-2
          Stem-loop Accession # MI0004806
          Stem-loop Sequence
          CUGUGGCUGGUAGGAUUCCUGGCAGAGUGAUUUGGAAAUGUAUGGUACAAAAUCACAUUGCCAGGGAUUUCCAAUCAGUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003568
          miRBase ID: xtr-miR-23a
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04270764_pri
          TaqMan™ Pri-miRNA Assay : Rn03465976_pri
          TaqMan™ Pri-miRNA Assay : Mm03306867_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10644
          Ambion® Pre-miR™ miRNA Precursor : PM10644
          TaqMan™ MicroRNA Assay : 000399
          mirVana® miRNA mimic : MC10644
          TaqMan™ Advanced miRNA Assay : 478532_mir
          TaqMan™ Advanced miRNA Assay : mmu478532_mir
          TaqMan™ Advanced miRNA Assay : rno478532_mir


          miRBase Alias:

          Human: hsa-miR-23a (v17)
          Mouse: mmu-miR-23a (v17)
          Rat: rno-miR-23a (v18)
          Aeromonas caviae Ae398: aca-miR-23a (v18)
          Macaca mulatta: mml-miR-23a (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline