Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13096
          Assay Name: hsa-miR-27a-5p
          miRBase Accession Number: MI0000085
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 13836440 - 13836517 [-] on Build GRCh38
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Pteropus alecto, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC13096

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000036, mir-27

          Mature miRNA Sequence:

          AGGGCUUAGCUGCUUGUGAGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-27a-5p MIMAT0004501
          Mouse mmu-miR-27a-5p MIMAT0004633
          Rat rno-miR-27a-5p MIMAT0004715
          Bovine bta-miR-27a-5p MIMAT0012532
          Cricetulus griseus cgr-miR-27a-5p MIMAT0023874
          Dasypus novemcinctus dno-miR-27a-5p MIMAT0047543
          View More Rows
          Goat chi-miR-27a-5p MIMAT0036105
          Guinea pig cpo-miR-27a-5p MIMAT0046890
          Pteropus alecto pal-miR-27a-5p MIMAT0039858
          Rhesus monkey mml-miR-27a-5p MIMAT0026600

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-27a
          Stem-loop Accession # MI0000085
          Stem-loop Sequence
          CUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUCCACACCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCAG
          Chromosome Location Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004501
          miRBase ID: hsa-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-27a
          Stem-loop Accession # MI0000578
          Stem-loop Sequence
          UGGCCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCCACAGCAAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGACCC
          Chromosome Location Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004633
          miRBase ID: mmu-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-27a
          Stem-loop Accession # MI0000860
          Stem-loop Sequence
          UGGCCUGUGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCUACAGCAAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGACCC
          Chromosome Location Chr. 19 - 25318736 - 25318822 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004715
          miRBase ID: rno-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: Chr. 19 - 25318736 - 25318822 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-27a
          Stem-loop Accession # MI0004746
          Stem-loop Sequence
          UGGCCUGGGGAGCAGGGCUUAGCUGCUUGUGAGCAGGUCCACAUCAAAUCGUGUUCACAGUGGCUAAGUUCCGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012532
          miRBase ID: bta-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-27a
          Stem-loop Accession # MI0020473
          Stem-loop Sequence
          CCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCCGCAGCAAGUCGUGUUCACAGUGGCUAAGUUCCGCUCCCCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023874
          miRBase ID: cgr-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-27a
          Stem-loop Accession # MI0038927
          Stem-loop Sequence
          AGGGCUUAGCUGCUUGUGAGCAGGGUCUGCAGCAAGUCGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047543
          miRBase ID: dno-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-27a
          Stem-loop Accession # MI0030722
          Stem-loop Sequence
          UGGCCUGGGGAGCAGGGCUUAGCUGCUUGUGAGCAGGUCCACAUCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036105
          miRBase ID: chi-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-27a
          Stem-loop Accession # MI0038583
          Stem-loop Sequence
          AGGGCUUAGCUGCUUGUGAGCAGGGUUCCCACCAGGCGUGUUCACAGUGGCUAAGUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046890
          miRBase ID: cpo-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-27a
          Stem-loop Accession # MI0032367
          Stem-loop Sequence
          AGGGCUUAGCUGCUUGUGAGCAGGGUCUGCACCAAGUCGUGUUCACAGUGGCUAAGUUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039858
          miRBase ID: pal-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-27a
          Stem-loop Accession # MI0003052
          Stem-loop Sequence
          CUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUCCACACCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026600
          miRBase ID: mml-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04270764_pri
          TaqMan™ Pri-miRNA Assay : Mm03306867_pri
          TaqMan™ Pri-miRNA Assay : Rn03465981_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13096
          Ambion® Pre-miR™ miRNA Precursor : PM13096
          TaqMan™ MicroRNA Assay : 002445
          mirVana® miRNA inhibitor : MH13096
          TaqMan™ Advanced miRNA Assay : rno481015_mir
          TaqMan™ Advanced miRNA Assay : 477998_mir
          TaqMan™ Advanced miRNA Assay : mmu481015_mir


          miRBase Alias:

          Human: hsa-miR-27a* (v17)
          Mouse: mmu-miR-27a* (v17)
          Rat: rno-miR-27a* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline