Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIM2 SIRNA Assays ›
          Gene Symbol:
          TRIM2
          Gene Name:
          tripartite motif containing 2
          Gene Aliases:
          CMT2R, RNF86
          Chromosome Location:
          Chr.4: 153152281 - 153339322 on Build GRCh38
          UniGene ID:
          Hs.435711
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 108691

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130067.1 NP_001123539.1 12 3506
          NM_015271.4 NP_056086.2 12 3503
          XM_006714157.1 XP_006714220.1 12 3515
          XM_006714158.1 XP_006714221.1 12 3480
          XM_006714159.3 XP_006714222.1 12 4326
          XM_006714160.1 XP_006714223.1 13 4280
          XM_006714161.1 XP_006714224.1 13 4277
          XM_006714162.1 XP_006714225.1 13 3997
          XM_006714165.3 XP_006714228.1 10 3708
          XM_011531794.1 XP_011530096.1 12 3594
          XM_011531795.1 XP_011530097.1 13 4000
          XM_011531796.2 XP_011530098.1 12 3530
          XM_011531798.2 XP_011530100.1 12 3635
          XM_017007943.1 XP_016863432.1 12 3468
          XM_017007944.1 XP_016863433.1 13 4268
          XM_017007945.1 XP_016863434.1 13 4265
          XM_017007946.1 XP_016863435.1 12 4311
          XM_017007947.1 XP_016863436.1 12 3582
          XM_017007948.1 XP_016863437.1 12 3536
          XM_017007949.1 XP_016863438.1 13 3985
          XM_017007950.1 XP_016863439.1 13 3988
          XM_017007951.1 XP_016863440.1 12 3682
          XM_017007952.1 XP_016863441.1 12 3690
          XM_017007953.1 XP_016863442.1 10 3046
          XM_017007954.1 XP_016863443.1 13 3560
          XM_017007955.1 XP_016863444.1 13 3553
          GenBank mRNA AB011089.1 - 12 3452
          AF220018.1 - 12 3452
          AL110234.1 - 5 1764
          BC011052.1 - 12 3506 4156234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          regulation of neuron apoptotic process
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0