Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 21 | 3213 | ||
NM_004505.3 | NP_004496.2 | 13 | 2682 | |||
XM_011524050.1 | XP_011522352.1 | 17 | 3431 | |||
XM_011524051.2 | XP_011522353.1 | 19 | 2526 | |||
XM_011524052.2 | XP_011522354.1 | 18 | 2640 | |||
XM_011524053.2 | XP_011522355.1 | 19 | 2553 | |||
XM_011524054.2 | XP_011522356.1 | 19 | 2520 | |||
XM_011524055.2 | XP_011522357.1 | 17 | 2447 | |||
XM_011524056.2 | XP_011522358.1 | 14 | 2400 | |||
XM_011524057.2 | XP_011522359.1 | 17 | 2563 | |||
XM_011524058.2 | XP_011522360.1 | 18 | 2629 | |||
XM_011524059.2 | XP_011522361.1 | 18 | 2629 | |||
XM_017025290.1 | XP_016880779.1 | 16 | 3269 | |||
GenBank mRNA | AB449915.1 | - | 12 | 983 | ||
AY143550.1 | - | 13 | 2682 | |||
BX647719.1 | - | 21 | 3214 | |||
X63546.1 | - | 13 | 2679 | |||
X63547.1 | - | 13 | 2679 |