Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242508.1 | NP_001229437.1 | 6 | 621 | ||
NM_018030.4 | NP_060500.3 | 4 | 990 | |||
NM_080597.3 | NP_542164.2 | 18 | 1877 | |||
XM_006722380.3 | XP_006722443.1 | 17 | 1790 | |||
XM_006722382.3 | XP_006722445.1 | 18 | 1890 | |||
XM_017025530.1 | XP_016881019.1 | 18 | 2169 | |||
XM_017025531.1 | XP_016881020.1 | 17 | 1662 | |||
XM_017025533.1 | XP_016881022.1 | 4 | 473 | |||
XM_017025534.1 | XP_016881023.1 | 4 | 3001 | |||
GenBank mRNA | AF274714.1 | - | 4 | 990 | ||
AF323726.2 | - | 18 | 1834 | |||
AF392449.1 | - | 4 | 611 | |||
AF392450.1 | - | 18 | 1834 | |||
AK001079.1 | - | 5 | 518 | |||
AK096358.1 | - | 18 | 1877 | |||
AK292895.1 | - | 19 | 1809 | |||
AK301862.1 | - | 6 | 621 | |||
AK314552.1 | - | 4 | 441 | |||
BC063420.1 | - | 4 | 676 | |||
BC073854.1 | - | 2 | 197 | |||
BC098384.1 | - | 4 | 669 | |||
BC136452.1 | - | 18 | 1774 | 9020441 | ||
BC144117.1 | - | 18 | 1774 | 9052637 | ||
BX647893.1 | - | 12 | 3553 |