Search
Search

WWP1| Gene Symbol: WWP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007013.3 | NP_008944.1 | 8 | 841 | ||
| XM_005250760.3 | XP_005250817.1 | 7 | 802 | |||
| XM_005250761.3 | XP_005250818.1 | 8 | 882 | |||
| XM_011516802.2 | XP_011515104.1 | 7 | 602 | |||
| XM_017012993.1 | XP_016868482.1 | 7 | 794 | |||
| XM_017012994.1 | XP_016868483.1 | 7 | 792 | |||
| XM_017012995.1 | XP_016868484.1 | 6 | 663 | |||
| XM_017012996.1 | XP_016868485.1 | 5 | 581 | |||
| XM_017012997.1 | XP_016868486.1 | 5 | 560 | |||
| GenBank mRNA | AK292800.1 | - | 8 | 856 | ||
| AL136739.1 | - | 6 | 571 | |||
| AY043361.1 | - | 7 | 646 | |||
| AY345857.1 | - | 8 | 875 | |||
| BC036065.1 | - | 8 | 845 | 5296005 | ||