Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP4K4 SIRNA Assays ›
          Gene Symbol:
          MAP4K4
          Gene Name:
          mitogen-activated protein kinase kinase kinase kinase 4
          Gene Aliases:
          FLH21957, HEL-S-31, HGK, MEKKK4, NIK
          Chromosome Location:
          Chr.2: 101697703 - 101894690 on Build GRCh38
          UniGene ID:
          Hs.701013
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1335

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAP4K4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242559.1 NP_001229488.1 4 202
          NM_001242560.1 NP_001229489.1 4 575
          NM_004834.4 NP_004825.3 4 575
          NM_145686.3 NP_663719.2 4 575
          NM_145687.3 NP_663720.1 4 202
          XM_005264048.2 XP_005264105.1 4 271
          XM_005264050.3 XP_005264107.1 4 271
          XM_005264052.3 XP_005264109.1 4 271
          XM_005264057.3 XP_005264114.1 4 271
          XM_005264058.3 XP_005264115.1 4 271
          XM_005264059.3 XP_005264116.1 4 271
          XM_005264060.3 XP_005264117.1 4 271
          XM_005264062.3 XP_005264119.1 4 271
          XM_005264063.3 XP_005264120.1 4 271
          XM_005264064.3 XP_005264121.1 4 271
          XM_005264065.3 XP_005264122.1 4 271
          XM_005264066.3 XP_005264123.1 4 271
          XM_005264068.3 XP_005264125.1 4 271
          XM_005264069.2 XP_005264126.1 4 271
          XM_005264071.2 XP_005264128.1 4 271
          XM_006712865.2 XP_006712928.1 4 271
          XM_006712868.2 XP_006712931.1 4 271
          XM_006712869.2 XP_006712932.1 4 271
          XM_017005349.1 XP_016860838.1 4 271
          XM_017005350.1 XP_016860839.1 4 271
          XM_017005351.1 XP_016860840.1 4 271
          XM_017005352.1 XP_016860841.1 4 271
          XM_017005353.1 XP_016860842.1 4 271
          XM_017005354.1 XP_016860843.1 4 271
          XM_017005355.1 XP_016860844.1 4 271
          XM_017005356.1 XP_016860845.1 4 271
          XM_017005357.1 XP_016860846.1 4 271
          XM_017005358.1 XP_016860847.1 4 271
          XM_017005359.1 XP_016860848.1 4 271
          XM_017005360.1 XP_016860849.1 4 271
          XM_017005361.1 XP_016860850.1 4 271
          XM_017005362.1 XP_016860851.1 4 271
          XM_017005363.1 XP_016860852.1 4 271
          XM_017005364.1 XP_016860853.1 4 271
          XM_017005365.1 XP_016860854.1 4 271
          XM_017005366.1 XP_016860855.1 4 271
          XM_017005367.1 XP_016860856.1 4 271
          XM_017005368.1 XP_016860857.1 4 271
          XM_017005369.1 XP_016860858.1 4 271
          XM_017005370.1 XP_016860859.1 4 271
          XM_017005371.1 XP_016860860.1 4 271
          XM_017005372.1 XP_016860861.1 4 271
          XM_017005373.1 XP_016860862.1 4 271
          XM_017005374.1 XP_016860863.1 4 271
          XM_017005375.1 XP_016860864.1 4 271
          GenBank mRNA AB014587.1 - 2 86
          AF096300.1 - 4 276
          AK295610.1 - 2 167
          AK296396.1 - 4 271
          AY212247.1 - 4 197
          AY834276.1 - 4 197
          DB086475.1 - 5 418
          EU794634.1 - 4 252

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106904, XM_008767010, XM_008767003, XM_008767007, XM_008767001, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008696, NM_001252200, NM_001252201, NM_001252202, BC141109, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          signal transduction by protein phosphorylation
          microvillus assembly
          intracellular signal transduction
          negative regulation of apoptotic process
          regulation of JNK cascade
          creatine kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline