Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSK3A SIRNA Assays ›
          Gene Symbol:
          GSK3A
          Gene Name:
          glycogen synthase kinase 3 alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 42230186 - 42243330 on Build GRCh38
          UniGene ID:
          Hs.466828
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1586

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GSK3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019884.2 NP_063937.2 2 574
          GenBank mRNA BC027984.1 - 2 593 3903896
          BC051865.2 - 2 563 6171438
          CR749620.1 - 1 1922
          D63424.1 - 2 546
          L40027.1 - 2 569

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure
          cardiac left ventricle morphogenesis
          glycogen metabolic process
          protein phosphorylation
          dopamine receptor signaling pathway
          nervous system development
          insulin receptor signaling pathway
          negative regulation of UDP-glucose catabolic process
          Wnt signaling pathway
          positive regulation of cAMP biosynthetic process
          negative regulation of TOR signaling
          cellular response to insulin stimulus
          cellular response to interleukin-3
          IRE1-mediated unfolded protein response
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of glycogen biosynthetic process
          positive regulation of protein catabolic process
          positive regulation of heart contraction
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          negative regulation of cell growth involved in cardiac muscle cell development
          positive regulation of adrenergic receptor signaling pathway
          negative regulation of canonical Wnt signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          chemical synaptic transmission, postsynaptic
          positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          regulation of mitophagy
          positive regulation of protein targeting to mitochondrion
          negative regulation of glycogen synthase activity, transferring glucose-1-phosphate
          negative regulation of type B pancreatic cell development
          negative regulation of glycogen (starch) synthase activity
          positive regulation of glycogen (starch) synthase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein kinase A catalytic subunit binding
          tau-protein kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0