Search
Search

Fads1| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146094.2 | NP_666206.1 | 2 | 473 | ||
| GenBank mRNA | AB072976.1 | - | 2 | 388 | ||
| AF061504.1 | - | 2 | 375 | |||
| AK080706.1 | - | 2 | 377 | |||
| AK083959.1 | - | 2 | 335 | |||
| AK154367.1 | - | 2 | 423 | |||
| BC022139.1 | - | 2 | 418 | 5100199 | ||
| BC026831.1 | - | 2 | 440 | 4239695 | ||
| BC026848.1 | - | 2 | 413 | 4196030 | ||
| BC063053.1 | - | 2 | 333 | 5696722 | ||
| KF987079.1 | - | 2 | 357 | |||