Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027652.2 | NP_081928.2 | 5 | 476 | ||
GenBank mRNA | AK014530.1 | - | 4 | 347 | ||
AK040745.1 | - | 5 | 385 | |||
AK048011.1 | - | 5 | 425 | |||
AK079325.1 | - | 5 | 452 | |||
AK122544.1 | - | 5 | 392 | |||
AK140416.1 | - | 4 | 339 | |||
BC106097.1 | - | 5 | 445 | 6485810 | ||
BC115783.1 | - | 5 | 415 | 40104916 | ||
BC117551.1 | - | 5 | 410 | 40104922 |