Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF206665.1 | - | 2 | 153 | ||
AF206666.1 | - | 1 | 98 | |||
BC051852.1 | - | 2 | 148 | |||
M30038.1 | - | 2 | 160 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 3 | 166 | ||
GenBank mRNA | AK308208.1 | - | 3 | 166 | ||
BC028059.1 | - | 2 | 155 | 5181815 | ||
M33493.1 | - | 2 | 120 |
Gene Symbol: TPSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012217.2 | NP_036349.1 | 2 | 173 | ||
GenBank mRNA | AF421357.1 | - | 1 | 47 | ||
BC069143.1 | - | 2 | 164 | 7217003 |