Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD44 SIRNA Assays ›
          Gene Symbol:
          CD44
          Gene Name:
          CD44 molecule (Indian blood group)
          Gene Aliases:
          CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
          Chromosome Location:
          Chr.11: 35138870 - 35232402 on Build GRCh38
          UniGene ID:
          Hs.502328
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 212873

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD44
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000610.3 NP_000601.3 18 2563
          NM_001001389.1 NP_001001389.1 17 2434
          NM_001001390.1 NP_001001390.1 12 1816
          NM_001001391.1 NP_001001391.1 9 1420
          NM_001001392.1 NP_001001392.1 4 800
          NM_001202555.1 NP_001189484.1 10 1624
          NM_001202556.1 NP_001189485.1 8 1357
          XM_005253231.2 XP_005253288.1 18 2293
          XM_005253232.2 XP_005253289.1 18 2290
          XM_005253233.2 XP_005253290.1 17 2173
          XM_005253234.2 XP_005253291.1 17 2170
          XM_005253235.2 XP_005253292.1 17 2164
          XM_005253238.2 XP_005253295.1 14 1804
          XM_005253239.2 XP_005253296.1 14 1798
          XM_005253240.2 XP_005253297.1 13 1669
          XM_006718388.1 XP_006718451.1 18 2287
          XM_011520482.1 XP_011518784.1 17 2089
          XM_011520483.1 XP_011518785.1 16 2038
          XM_011520484.1 XP_011518786.1 16 2035
          XM_011520485.1 XP_011518787.1 16 2032
          XM_011520486.1 XP_011518788.1 13 1675
          XM_011520488.1 XP_011518790.1 11 1339
          XM_017018583.1 XP_016874072.1 17 2161
          XM_017018584.1 XP_016874073.1 11 1402
          XM_017018585.1 XP_016874074.1 10 1273
          GenBank mRNA AF098641.1 - 4 384
          AJ251595.1 - 18 2307
          AK129808.1 - 8 815
          AK290424.1 - 9 986
          AK297776.1 - 9 1026
          AK304467.1 - 8 923
          AK307707.1 - 8 881
          AY101192.1 - 12 1498
          AY101193.1 - 9 1103
          BC004372.1 - 17 2131 3638681
          BC052287.1 - 8 974
          BC067348.1 - 9 1103
          EF581837.1 - 10 1201
          FJ216964.1 - 8 923
          M24915.1 - 9 1101
          M59040.1 - 9 1014
          S66400.1 - 12 1432
          U40373.1 - 9 1050
          W47313.1 - 2 134
          X55150.1 - 12 1505
          X56794.1 - 12 1497
          X66733.1 - 17 2129

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012924, XM_008762056, XM_008762057, XM_008762059, XM_006234627, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001039151, NM_001177787, NM_001177785, NM_001039150, NM_009851, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          single organismal cell-cell adhesion
          extracellular matrix disassembly
          extracellular matrix organization
          hyaluronan catabolic process
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of heterotypic cell-cell adhesion
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          cellular response to fibroblast growth factor stimulus
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          cartilage development
          interferon-gamma-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          monocyte aggregation
          positive regulation of monocyte aggregation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          cytokine receptor activity
          protein binding
          collagen binding
          hyaluronic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xhspb:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0