Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK10 SIRNA Assays ›
Gene Symbol:
NEK10
Gene Name:
NIMA related kinase 10
Gene Aliases:
-
Chromosome Location:
Chr.3: 27107336 - 27369460 on Build GRCh38
UniGene ID:
Hs.506115
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
213347
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152534.4
NP_689747.3
3
334
NM_199347.3
NP_955379.2
3
334
XM_006712997.3
XP_006713060.1
3
295
XM_006712998.2
XP_006713061.1
3
295
XM_006712999.3
XP_006713062.1
3
295
XM_006713001.3
XP_006713064.1
3
295
XM_006713002.1
XP_006713065.1
3
295
XM_011533413.2
XP_011531715.1
3
295
XM_011533414.2
XP_011531716.1
3
295
XM_011533415.1
XP_011531717.1
3
295
XM_017005761.1
XP_016861250.1
2
247
XM_017005762.1
XP_016861251.1
4
411
XM_017005763.1
XP_016861252.1
3
392
XM_017005764.1
XP_016861253.1
2
224
XM_017005765.1
XP_016861254.1
2
196
XM_017005766.1
XP_016861255.1
2
247
XM_017005767.1
XP_016861256.1
2
247
XM_017005768.1
XP_016861257.1
3
295
XM_017005769.1
XP_016861258.1
3
295
XM_017005770.1
XP_016861259.1
2
247
XM_017005771.1
XP_016861260.1
2
247
XM_017005772.1
XP_016861261.1
2
247
XM_017005773.1
XP_016861262.1
3
295
XM_017005774.1
XP_016861263.1
2
247
GenBank mRNA
AK123061.1
-
3
295
AK128585.1
-
2
224
DB094082.1
-
2
295
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
microtubule-based process
chromosome segregation
positive regulation of protein autophosphorylation
positive regulation of MAP kinase activity
regulation of ERK1 and ERK2 cascade
regulation of cell cycle G2/M phase transition
protein kinase activity
protein serine/threonine kinase activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0