Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTLL9 SIRNA Assays ›
Gene Symbol:
TTLL9
Gene Name:
tubulin tyrosine ligase like 9
Gene Aliases:
C20orf125
Chromosome Location:
Chr.20: 31870702 - 31944959 on Build GRCh38
UniGene ID:
Hs.712915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
218679
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTLL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008409.2
NP_001008409.1
5
530
NR_134519.1
6
616
XM_005260309.2
XP_005260366.1
4
294
XM_006723723.2
XP_006723786.1
3
231
XM_006723724.2
XP_006723787.1
4
403
XM_011528617.1
XP_011526919.1
5
366
XM_011528618.1
XP_011526920.1
5
530
XM_011528619.1
XP_011526921.1
6
428
XM_011528620.2
XP_011526922.1
6
425
XM_011528621.1
XP_011526923.1
6
443
XM_011528622.1
XP_011526924.1
6
418
XM_011528623.2
XP_011526925.1
4
299
XM_011528624.1
XP_011526926.1
5
366
XM_011528625.1
XP_011526927.1
5
366
XM_011528626.1
XP_011526928.1
5
364
XM_011528627.2
XP_011526929.1
5
361
XM_011528628.1
XP_011526930.1
4
289
XM_011528629.1
XP_011526931.1
4
297
XM_011528630.1
XP_011526932.1
3
231
XM_011528631.1
XP_011526933.1
5
366
XM_011528632.1
XP_011526934.1
5
366
XM_011528633.1
XP_011526935.1
5
366
XM_011528634.1
XP_011526936.1
5
366
XM_011528635.1
XP_011526937.1
5
366
XM_011528637.2
XP_011526939.1
5
366
XM_011528638.1
XP_011526940.1
5
366
XM_011528639.1
XP_011526941.1
5
366
XM_011528640.1
XP_011526942.1
5
366
XM_011528641.1
XP_011526943.1
5
366
XM_011528643.2
XP_011526945.1
5
366
XM_011528644.1
XP_011526946.1
5
366
XM_011528646.1
XP_011526948.1
6
555
XM_017027694.1
XP_016883183.1
3
231
XM_017027695.1
XP_016883184.1
5
366
XM_017027696.1
XP_016883185.1
5
366
XM_017027697.1
XP_016883186.1
5
366
XM_017027698.1
XP_016883187.1
5
366
XM_017027699.1
XP_016883188.1
4
549
XM_017027700.1
XP_016883189.1
5
366
XM_017027701.1
XP_016883190.1
5
366
XM_017027702.1
XP_016883191.1
5
366
XM_017027703.1
XP_016883192.1
5
366
GenBank mRNA
AK093491.1
-
5
531
AK302360.1
-
3
233
AK302806.1
-
4
277
BC068593.1
-
6
607
BC104024.1
-
6
555
40025306
BC104025.1
-
4
403
40025308
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0