Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PMS2P1 SIRNA Assays ›
          Gene Symbol:
          PMS2P1 DTX2P1-UPK3BP1-PMS2P11 PMS2P2 PMS2P4 PMS2P5 PMS2P9 POLR2J3
          Gene Name:
          PMS1 homolog 2, mismatch repair system component pseudogene 1
          DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
          PMS1 homolog 2, mismatch repair system component pseudogene 2
          PMS1 homolog 2, mismatch repair system component pseudogene 4
          PMS1 homolog 2, mismatch repair system component pseudogene 5
          PMS1 homolog 2, mismatch repair system component pseudogene 9
          RNA polymerase II subunit J3
          Gene Aliases:
          PMS2L1, PMS2L13, PMS2L6, PMS2L7, PMS2L8, PMS3, PMS8, PMSR1, PMSR2
          PMS2L11
          PMS2L2, PMS4
          PMS2L4, PMS6
          PMS2L5, PMS7
          PMS2L17, PMS2LP1, PMSR5
          POLR2J2, RPB11b1, RPB11b2
          Chromosome Location:
          Chr.7: 100320640 - 100336307 on Build GRCh38
          Chromosome Location:
          Chr.7: 76980822 - 77023761 on Build GRCh38
          Chromosome Location:
          Chr.7: 75343937 - 75358997 on Build GRCh38
          Chromosome Location:
          Chr.7: 67276131 - 67302442 on Build GRCh38
          Chromosome Location:
          Chr.7: 74890768 - 74921138 on Build GRCh38
          Chromosome Location:
          Chr.7: 77039480 - 77053038 on Build GRCh38
          Chromosome Location:
          Chr.7: 102537919 - 102572660 on Build GRCh38
          UniGene ID:
          Hs.634244
          Hs.675888
          Hs.292996
          Hs.712714
          Hs.292996
          Hs.292996
          Hs.610255
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 279266

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PMS2P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003613.1 7 1164
          GenBank mRNA AB017004.1 - 7 1164
          BC067809.1 - 6 707
          D38435.1 - 7 767
          D38499.1 - 7 767
          DA706474.1 - 2 210
          Gene Symbol: DTX2P1-UPK3BP1-PMS2P11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_023383.1 11 1686
          Gene Symbol: PMS2P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003614.3 5 558
          Gene Symbol: PMS2P4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046297.1 6 664
          Gene Symbol: PMS2P5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027775.2 7 773
          NR_027776.2 6 670
          GenBank mRNA AK309660.1 - 5 529
          BF984173.1 - 6 672
          D38436.1 - 7 754
          D38502.1 - 7 754
          Gene Symbol: PMS2P9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028058.1 5 1070
          Gene Symbol: POLR2J3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DA580738.1 - 5 518

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          phosphorylation
          somatic hypermutation of immunoglobulin genes
          single-stranded DNA binding
          kinase activity
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0