Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LYRM1 SIRNA Assays ›
Gene Symbol:
LYRM1
Gene Name:
LYR motif containing 1
Gene Aliases:
A211C6.1
Chromosome Location:
Chr.16: 20899626 - 20925006 on Build GRCh38
UniGene ID:
Hs.745012
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28588
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LYRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128301.2
NP_001121773.1
4, 5
712
NM_001128302.2
NP_001121774.1
3, 4
480
NM_001302835.1
NP_001289764.1
5, 6
656
NM_001302836.1
NP_001289765.1
5, 6
808
NM_020424.4
NP_065157.1
4, 5
631
NR_126516.1
3, 4
553
NR_126520.1
2, 3
335
NR_126521.1
2, 3
321
NR_126527.1
2, 3
149
NR_126528.1
3, 4
567
XM_005255444.1
XP_005255501.1
4, 5
533
XM_011545900.2
XP_011544202.1
4, 5
295
XM_011545901.2
XP_011544203.1
4, 5
361
XM_011545902.1
XP_011544204.1
5, 6
632
XM_011545908.2
XP_011544210.1
4, 5
363
XM_017023475.1
XP_016878964.1
5, 6
1048
XM_017023477.1
XP_016878966.1
3, 4
883
XM_017023478.1
XP_016878967.1
4, 5
353
XM_017023479.1
XP_016878968.1
4, 5
790
XM_017023480.1
XP_016878969.1
6, 7
876
XM_017023481.1
XP_016878970.1
4, 5
505
XM_017023482.1
XP_016878971.1
3, 4
559
GenBank mRNA
AK308690.1
-
2, 3
400
AK311415.1
-
3, 4
555
AK311881.1
-
3, 4
347
AV714321.1
-
3, 4
326
BC017039.1
-
5, 6
656
3849248
BF575136.1
-
3, 4
369
BG541469.1
-
2, 3
184
BI547471.1
-
3, 4
398
BI825882.1
-
5, 6
655
BM546065.1
-
2, 3
285
BX096767.1
-
2, 3
149
CD237780.1
-
3, 4
350
CX163936.1
-
3, 4
357
DA572182.1
-
4, 5
597
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0