Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207348.2 | NP_997231.1 | 5 | 1724 | ||
XM_011541292.2 | XP_011539594.1 | 6 | 3024 | |||
XM_017001083.1 | XP_016856572.1 | 4 | 2815 | |||
GenBank mRNA | AK024244.1 | - | 1 | 445 | ||
AL832282.1 | - | 1 | 799 | |||
BC027998.1 | - | 5 | 1724 | 5189944 | ||
CR749264.1 | - | 2 | 961 |