Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GOSR2 SIRNA Assays ›
          Gene Symbol:
          GOSR2
          Gene Name:
          golgi SNAP receptor complex member 2
          Gene Aliases:
          Bos1, EPM6, GS27
          Chromosome Location:
          Chr.17: 46923086 - 46975519 on Build GRCh38
          UniGene ID:
          Hs.463278
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 35686

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GOSR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012511.2 NP_001012529.1 3, 4 272
          NM_001321133.1 NP_001308062.1 3, 4 272
          NM_001321134.1 NP_001308063.1 3, 4 320
          NM_004287.4 NP_004278.2 3, 4 272
          NM_054022.3 NP_473363.1 3, 4 272
          XM_005257844.3 XP_005257901.1 3, 4 281
          XM_006722190.3 XP_006722253.1 3, 4 281
          XM_011525501.2 XP_011523803.1 3, 4 281
          XM_011525502.2 XP_011523804.1 3, 4 281
          XM_017025378.1 XP_016880867.1 3, 4 278
          XM_017025379.1 XP_016880868.1 3, 4 272
          XM_017025380.1 XP_016880869.1 3, 4 629
          XM_017025381.1 XP_016880870.1 3, 4 255
          XM_017025382.1 XP_016880871.1 3, 4 607
          XM_017025383.1 XP_016880872.1 3, 4 281
          XM_017025384.1 XP_016880873.1 3, 4 303
          XM_017025385.1 XP_016880874.1 3, 4 278
          XM_017025386.1 XP_016880875.1 3, 4 281
          XM_017025387.1 XP_016880876.1 3, 4 278
          XM_017025388.1 XP_016880877.1 3, 4 281
          XM_017025389.1 XP_016880878.1 3, 4 288
          XM_017025390.1 XP_016880879.1 3, 4 633
          XM_017025391.1 XP_016880880.1 3, 4 631
          XM_017025392.1 XP_016880881.1 3, 4 281
          XM_017025393.1 XP_016880882.1 3, 4 278
          XM_017025394.1 XP_016880883.1 3, 4 629
          XM_017025395.1 XP_016880884.1 3, 4 629
          GenBank mRNA AF007548.1 - 3, 4 196
          AF229796.1 - 3, 4 227
          AK290890.1 - 3, 4 247
          BC009710.2 - 3, 4 243 3890308
          BC034762.1 - 3, 4 215 5185659
          BM846811.1 - 3, 4 266
          CD048223.1 - 3, 4 297

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to vacuole
          ER to Golgi vesicle-mediated transport
          intra-Golgi vesicle-mediated transport
          Golgi to vacuole transport
          IRE1-mediated unfolded protein response
          retrograde transport, endosome to Golgi
          COPII vesicle coating
          vesicle fusion with Golgi apparatus
          membrane fusion
          SNARE binding
          transporter activity
          SNAP receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0