Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HRAS SIRNA Assays ›
          Gene Symbol:
          HRAS
          Gene Name:
          HRas proto-oncogene, GTPase
          Gene Aliases:
          C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, RASH1, p21ras
          Chromosome Location:
          Chr.11: 532242 - 535567 on Build GRCh38
          UniGene ID:
          Hs.37003
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42267

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HRAS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130442.2 NP_001123914.1 3 477
          NM_001318054.1 NP_001304983.1 3 477
          NM_005343.3 NP_005334.1 3 477
          NM_176795.4 NP_789765.1 3 477
          GenBank mRNA AB451336.1 - 2 272
          AB451485.1 - 2 272
          AF493916.1 - 2 272
          AJ437024.1 - 2 272
          BC006499.2 - 3 448 2819996
          BC095471.1 - 2 273 6519193
          BM801600.1 - 3 400
          BM801632.1 - 3 460
          BM808879.1 - 485
          BQ674260.1 - 2 335
          BT019421.1 - 2 272
          CR536579.1 - 2 272
          CR542271.1 - 2 272
          KJ534867.1 - 2 272
          KJ534868.1 - 2 272

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          positive regulation of protein phosphorylation
          stimulatory C-type lectin receptor signaling pathway
          endocytosis
          chemotaxis
          cell cycle arrest
          mitotic cell cycle checkpoint
          signal transduction
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          Ras protein signal transduction
          axon guidance
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          organ morphogenesis
          negative regulation of gene expression
          positive regulation of cell migration
          positive regulation of interferon-gamma production
          negative regulation of GTPase activity
          social behavior
          Fc-epsilon receptor signaling pathway
          ERBB2 signaling pathway
          T-helper 1 type immune response
          defense response to protozoan
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          positive regulation of DNA replication
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of Ras protein signal transduction
          ephrin receptor signaling pathway
          regulation of long-term neuronal synaptic plasticity
          positive regulation of epithelial cell proliferation
          T cell receptor signaling pathway
          leukocyte migration
          protein heterooligomerization
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of wound healing
          cellular senescence
          intrinsic apoptotic signaling pathway
          positive regulation of ruffle assembly
          positive regulation of actin cytoskeleton reorganization
          positive regulation of miRNA metabolic process
          GTPase activity
          protein binding
          GTP binding
          protein C-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline