Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307960.1 | NP_001294889.1 | 4 | 507 | ||
NM_001308026.1 | NP_001294955.1 | 5 | 585 | |||
NM_025141.3 | NP_079417.2 | 4 | 507 | |||
NM_078474.2 | NP_510883.2 | 5 | 585 | |||
GenBank mRNA | AF353992.1 | - | 4 | 482 | ||
AK026257.1 | - | 4 | 497 | |||
AK094955.1 | - | 3 | 917 | |||
AK297048.1 | - | 4 | 492 | |||
AK315584.1 | - | 5 | 561 | |||
AL512689.1 | - | 1 | 1343 | |||
BC006150.1 | - | 5 | 557 | 3051281 | ||
BC008873.2 | - | 4 | 480 | 4137334 | ||
BU595412.1 | - | 5 | 580 |