siRNA Details
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 2 | 1269 | ||
NM_018703.3 | NP_061173.1 | 2 | 1269 | |||
NM_032626.5 | NP_116015.2 | 2 | 1269 | |||
GenBank mRNA | AB112074.1 | - | 2 | 273 | ||
AB112075.1 | - | 2 | 273 | |||
BC029352.1 | - | 2 | 360 | |||
BC063524.1 | - | 2 | 336 | |||
BC101139.2 | - | 2 | 391 | |||
BC101140.2 | - | 2 | 391 | |||
BC101141.2 | - | 2 | 391 | |||
BC101142.1 | - | 2 | 391 | |||
BC114353.1 | - | 2 | 391 | |||
BC114354.1 | - | 2 | 391 | |||
BC118667.1 | - | 2 | 391 | |||
BC139830.1 | - | 2 | 391 | |||
BX648661.1 | - | 2 | 209 | |||
CD638608.1 | - | 2 | 123 |

More Information
Other Species Targeted

Panther Classification:
